
Cyanine 3 Phosphoramidite

Synonym(s):
2-[3-[1,3-Dihydro-1-[3-(4-methoxytriphenylmethoxy)propyl]-3,3-dimethyl-2H-indol-2-ylidene]-1-propen-1-yl]-1-[3-[N,N-diisopropylamino(2-cyanoethoxy)phosphinoxy]propyl]-3,3-dimethyl-3H-indolium
Empirical Formula (Hill Notation):
C58H70N4O4P
CAS Number:
Molecular Weight:
918.17
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Product Line
Quality Segment
Technique
Showing 1-3 of 3