
Cefoxitin sodium

Synonym(s):
Cefoxitin sodium salt
Empirical Formula (Hill Notation):
C16H16N3NaO7S2
CAS Number:
Molecular Weight:
449.43
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing