
Bisacodyl for peak identification

Synonym(s):
Bisacodyl
Empirical Formula (Hill Notation):
C22H19NO4
CAS Number:
Molecular Weight:
361.39
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing