
Atractylenolide I

Empirical Formula (Hill Notation):
C15H18O2
CAS Number:
Molecular Weight:
230.30
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing