
Arg-Arg β-naphthylamide trihydrochloride

Empirical Formula (Hill Notation):
C22H33N9O2 · 3HCl
CAS Number:
Molecular Weight:
564.94
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing