
7-Methoxycoumarin

Synonym(s):
Herniarin, Methyl umbelliferyl ether
Empirical Formula (Hill Notation):
C10H8O3
CAS Number:
Molecular Weight:
176.17
Products
Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Organoleptic Class
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing