
3,5-Dichlorophenyldiazonium tetrafluoroborate

Empirical Formula (Hill Notation):
C6H3Cl2N2 · BF4
CAS Number:
Molecular Weight:
260.81
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing