
3,3′-Diaminobenzidine tetrahydrochloride

Synonym(s):
3,3′,4,4′-Biphenyltetramine tetrahydrochloride, 3,3′,4,4′-Tetraaminobiphenyl tetrahydrochloride, DAB
Linear Formula:
(NH2)2C6H3C6H3(NH2)2 · 4HCl
Molecular Weight:
360.11
EC Number:
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing