Skip to Content
Merck
All Photos(1)

Key Documents

EP5810709008

Eppendorf® Swing-bucket rotor A-4-62

incl. 4 x 250 ml rectangular buckets

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103909
NACRES:
NB.82

feature

to hold 4 x 250 mL rectangular buckets
incl. 4 x 250 ml rectangular buckets

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 5810709008

parameter

4,000 max. RCF (3,220 xg)

technique(s)

centrifugation: suitable

compatibility

for use with centrifuge models 5804/5804R/5810/5810R

General description

This rotor can be used for a wide range of molecular applications. Eppendorf Swing Bucket Rotors are designed for use with Eppendorf benchtop centrifuges.

Features and Benefits

  • Accommodates tubes and plates
  • Aerosol-tight caps for safe centrifugation of hazardous samples optionally available
  • Rotor life time: max. 40,000 cycles
  • Rotor, buckets, caps and adapters are autoclavable

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service