Skip to Content
Merck
All Photos(1)

Key Documents

Y0001554

Ciclesonide containing impurity A

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

biological source

synthetic

grade

pharmaceutical primary standard

Agency

EP

API family

ciclesonide

packaging

pkg of 15 mg

manufacturer/tradename

EDQM

storage condition

protect from light

color

white

bp

665.0 °C/1.333 hPa (1229.0°F)

mp

209-211 °C (408—412°F)

solubility

water: <0.1 g/L
acetone: soluble
ethanol: soluble

density

1.23 g/cm3 at 20 °C (1.333 hPa)

application(s)

pharmaceutical (small molecule)

format

neat

shipped in

ambient

storage temp.

2-8°C

General description

Ciclesonide is a new-generation, non-halogenated glucocorticoid. It is a prodrug that releases an active primary metabolite, ciclesonide−active principle (CIC-AP), on enzymatic cleavage.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ciclesonide containing impurity A is used as an EP reference standard to quantify the analyte in pharmaceutical formulations by liquid chromatography (LC) technique.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Population pharmacokinetics and pharmacodynamics of ciclesonide
Rohatagi S, et al.
Journal of Clinical Pharmacology, 43(4), 365-378 (2003)
New Drugs for Asthma, Allergy and COPD, 42, 42-54 (2001)
Rethinking Cleaning Validation for API Manufacturing
Zhang C, et al.
Pharmaceutical Technology, 42, 42-54 (2018)
Pharmacokinetics of [14 C] ciclesonide after oral and intravenous administration to healthy subjects
Nave R, et al.
Clinical Pharmacokinetics, 43, 479-486 (2004)
Ciclesonide
European Pharmacopoeia Commission and European Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 43, 2206-2207 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service