Skip to Content
Merck
All Photos(3)

Key Documents

T3163

Sigma-Aldrich

Tetramethylrhodamine isothiocyanate Isomer R

powder

Synonym(s):

6-TRITC, Tetramethylrhodamine-6-isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H21N3O3S
CAS Number:
Molecular Weight:
443.52
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

description

labeling efficiency with bovine albumin >= 50%

Quality Level

form

powder

composition

Dye content, ≥80%

solubility

1 M NH4OH: 10 mg/mL (Faint Red to Very Dark Red to Very Dark Purple solution)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

CN(C)c1ccc2c(OC3=C\C(C=CC3=C2c4cc(ccc4C([O-])=O)N=C=S)=[N+](/C)C)c1

InChI

1S/C25H21N3O3S/c1-27(2)16-6-9-19-22(12-16)31-23-13-17(28(3)4)7-10-20(23)24(19)21-11-15(26-14-32)5-8-18(21)25(29)30/h5-13H,1-4H3

InChI key

OBYNJKLOYWCXEP-UHFFFAOYSA-N

General description

Tetramethylrhodamine isothiocyanate Isomer R (TRITC) is the R isomer of tetramethylrhodamine isothiocyanate and is also known as 6-tetramethylrhodamine isothiocyanate. It is a strongly fluorescent compound soluble in ethanol, methanol, dimethylformamide, dimethylsulfoxide, and water. TRITC is a moderately-sized aminoxanthene dye carrying carboxylic acid and isothiocyanate substituents on the non-planar pendant phenyl ring. It forms a lipophilic cation in acidic pH and a hydrophilic zwitterion around neutral pH. The isothiocyanate group reacts with nucleophilic substituents (e.g., amino, hydroxyl, thiol) of biomolecules, providing the means of attaching a fluorescent label. TRITC also reacts with water and hydroxide ions and is unstable in aqueous solutions. TRITC is commercially available as a zwitterion or as a chloride.

Application

  • TRITC is used to label a wide variety of biomolecules, including immunoglobulins, lectins, nucleic acids, polynucleotides, and polysaccharides for affinity, immuno-, and in situ hybridization, fluorescent probes, and flow cytometry.
  • Lectins are TRITC-Iabeled reagents for the affinity staining of sections and cell monolayers to distinguish the sialomucins of salivary glands.
  • TRITC-antibodies are used to identify pathogenic amoebae.
  • TRITC-oligonucleotides are used in the in-situ hybridization staining of soil microorganisms.
  • TRITC-Iabeled reagents have also been used as fluorescent probes of live cells.
  • Flow cytometry of TRlTC-Iabeled slime mold cells has been used to study their aggregation behavior.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In Vitro Study of Nuclear Assembly and Nuclear
Import Using Xenopus Egg Extracts
Douglass J Forbes
Methods in Molecular Biology (2016)
Conn's Biological Stains
Kiernan & Horobin
Conn?s Biological Stains (2002)
E. Harlow and D. Lane, ed.
Antibodies: A Laboratory Manual, 353-355 (1988)
Yanyan Jia et al.
Neuron, 105(2), 310-321 (2019-11-26)
Transmembrane channel-like (TMC) 1 and 2 are required for the mechanotransduction of mouse inner ear hair cells and localize to the site of mechanotransduction in mouse hair cell stereocilia. However, it remains unclear whether TMC1 and TMC2 are indeed ion
A simple method for determining the labeling efficiency of fluorescein isothiocyanate products.
R M McKinney et al.
Analytical biochemistry, 14(3), 421-428 (1966-03-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service