Skip to Content
Merck
All Photos(2)

Key Documents

32486

Supelco

Spirotetramat Metabolite BYI08330-mono-hydroxy

PESTANAL®, analytical standard

Synonym(s):

cis-3-(2,5-Dimethylphenyl)-4-hydroxy-8-methoxy-1-azaspiro[4.5]decan-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H25NO3
CAS Number:
Molecular Weight:
303.40
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

−20°C

SMILES string

COC1CCC2(CC1)NC(=O)C(C2O)c3cc(C)ccc3C

InChI

1S/C18H25NO3/c1-11-4-5-12(2)14(10-11)15-16(20)18(19-17(15)21)8-6-13(22-3)7-9-18/h4-5,10,13,15-16,20H,6-9H2,1-3H3,(H,19,21)/t13-,15?,16?,18+

InChI key

HPQGJNTUXNUIDL-RMVSHPHESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Spirotetramat is the member of cyclic ketoenoles. It is an inhibitor of acetyl-CoA carboxylase which is active against a wide spectrum of sucking insects.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemometric-assisted QuEChERS extraction method for the residual analysis of thiacloprid, spirotetramat and spirotetramat?s four metabolites in pepper: application of their dissipation patterns.
Li S, et al.
Food Chemistry, 192, 893-899 (2016)
Simultaneous determination of spirotetramat and its four metabolites in fruits and vegetables using a modified quick, easy, cheap, effective, rugged, and safe method and liquid chromatography/tandem mass spectrometry.
Zhu Y, et al.
Journal of Chromatography A, 1299, 71-77 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service