Skip to Content
Merck
All Photos(1)

Key Documents

EZSPKIT01

Millipore

EZ-Pak® Membrane Dispenser Spare Part Kit

Spare parts for use with EZ-Pak® Membrane Dispenser

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

Agency

USP 71

Quality Level

manufacturer/tradename

EZ-Pak®

input

sample type beverage(s)
sample type pharmaceutical(s)
sample type food(s)

application(s)

bioburden testing
food and beverages
pharmaceutical

compatibility

for use with EZ-Pak® Dispenser Curve

shipped in

ambient

General description

The Spare part kit is an additional accessory of the EZ-Pak® Dispenser. The EZ-Pak® Dispenser Curve revolutionizes filter handling for membrane filtration method testing by combining the benefits of high-speed sterile membrane dispensing, no-touch operation, and one-handed membrane transfer. The spare parts aid in easy membrane loading and dispensing.

Application

The Spare part kit is an additional accessory of the EZ-Pak® Dispenser Curve used in general microbiological analysis of Beer, Bottled Water, Pharmaceutical Products Analysis, Environmental Analysis, Bioburden Testing, and Water Monitoring.

Components

  • 3 actuators
  • 1 clip
  • 4 feet
  • 2 sets of latch springs
  • 2 sets of driving belts
  • 2,5mm and 3mm Allen keys
  • latch spring installation tool
  • latch
  • cover attaching screws
  • 2 tensioning springs

Legal Information

EZ-PAK is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service