Skip to Content
Merck
All Photos(1)

Key Documents

444294-M

Millipore

MMP-2 Inhibitor IV

The MMP-2 Inhibitor IV controls the biological activity of MMP-2. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

MMP-2 Inhibitor IV, (4′-chlorobiphenyl-4-ylsulfonamido)methylenediphosphonic acid, Bone Resorption Inhibitor, Bone Resorption Inhibitor, (4′-chlorobiphenyl-4-ylsulfonamido)methylenediphosphonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H14ClNO8P2S
Molecular Weight:
441.72
UNSPSC Code:
12352200

Quality Level

Assay

≥99% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL, clear, colorless

storage temp.

2-8°C

General description

A cell-permeable, bisphosphonate derivative that displays nanomolar activity against MMP-2 (IC50 = 37 nM), with good selectivity over MMP-8 (320 nM), MMP-9 (>1 M), and MMP-14 (>1 M). It is shown to inhibit osteoclast activity in a macrophage cell line, J774 (IC50 = 1.7 M), which is more potent compared with other bisphosphonates such as Alendronate (Cat. No. 126855), (IC50 = 30 M), and Zolendronate (7.8 M), without observable cytotoxicity in HepG2 cells. At 25 M, it abolishes the formation of actin rings, which are functional structures that are typical of resorbing osteoclasts, at activity levels comparable with those of zolendronic acid, inhibits bone resportion in vitro, and demonstrates cytotoxic properties in murine osteoclasts.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Rubino, M.T., et al. 2011. Chem. Med. Chem.6, 1258.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service