Skip to Content
Merck
All Photos(1)

Key Documents

444286-M

Sigma-Aldrich

MMP-2 Inhibitor II

The MMP-2 Inhibitor II, also referenced under CAS 869577-51-5, controls the biological activity of MMP-2. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

MMP-2 Inhibitor II, (4-(4-(Methanesulfonamido)phenoxy)phenylsulfonyl)methyloxirane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H17NO6S2
CAS Number:
Molecular Weight:
383.44
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

solid

potency

2.4 μM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

color

white

solubility

DMSO: 200 mg/mL

storage temp.

−20°C

InChI

1S/C16H17NO6S2/c1-24(18,19)17-12-2-4-13(5-3-12)23-14-6-8-16(9-7-14)25(20,21)11-15-10-22-15/h2-9,15,17H,10-11H2,1H3

InChI key

ADEXXMLIERNFQB-UHFFFAOYSA-N

General description

An oxirane analog of SB-3CT, pMS (Cat. No. 444285) that acts as a selective, active site-binding, irreversible inhibitor of MMP-2 (Ki = 2.4 µM). Although less potent, it exhibits enhanced selectivity towards MMP-2 (Ki = 45 and 379 µM for MMP-1 and MMP-7, respectively) than SB-3CT, pMS.

Biochem/physiol Actions

Secondary Target
MMP-2 (Ki = 45 and 379 µM for MMP-1 and MMP-7, respectively)

Warning

Toxicity: Standard Handling (A)

Other Notes

Ikejiri, M., et al. 2005. J. Biol. Chem.280, 33992.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service