Skip to Content
Merck
All Photos(1)

Key Documents

14-919-M

Sigma-Aldrich

LRRK2 Protein, active, 10 µg

Active, N-terminal FLAG-tagged, recombinant, human LRRK2, amino acids 970-end. For use in Kinase Assays.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

biological source

human

Quality Level

100
300

recombinant

expressed in baculovirus infected Sf21 cells

mol wt

Mw 180 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

GenBank accession no.

UniProt accession no.

Gene Information

human ... LRRK2(120892)

General description

Product Source: N-terminal FLAG-tagged, recombinant, human LRRK2 amino acids 970-end, expressed by baculovirus in Sf21 insect cells.
This protein is a member of the leucine-rich repeat kinase family and encodes a protein with an ankryin repeat region, a leucine-rich repeat (LRR) domain, a kinase domain, a DFG-like motif, a RAS domain, a GTPase domain, a MLK-like domain, and a WD40 domain. The protein is present largely in the cytoplasm but also associates with the mitochondrial outer membrane. Mutations in this gene have been associated with Parkinson disease-8.

Packaging

Also available in 250μg size as catalog # 14-919M and in bulk as catalog # 14-919-K.

Quality

Routinely evaluated by phosphorylation of ERMtide (RLGRDKYKTLRQIRQ)

Physical form

50mM Tris/HCl pH7.4, 300mM NaCl, 20mM β-glycerophosphate, 0.25mM sodium orthovanadate, 10mM NaF, 150µg/ml FLAG peptide, 20% v/v glycerol, 1mM benzamidine, 0.2mM PMSF, 5mM DTT. Frozen solution.

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service