SLW1000/38D
Pyrex® Griffin beakers, low form, with printed trace code
10000 mL
Synonym(s):
Pyrex, SciLabware 1000/38D
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
white graduation
feature
low form
with printed trace code
manufacturer/tradename
1000/38D
SciLabware
H × diam.
350 mm × 217 mm
capacity
10000 mL
General description
- Complies with ISO 3819 and DIN 12331
- Manufactured from Pyrex borosilicate glass
- Excellent thermal performance
- High resistance to chemical attack
- Printed with Trace Code for downloadable batch certificate
- Batch certificate available to download from www.scilabware.com
- White graduations and marking spot
Other Notes
Browse all glass beakers
Legal Information
PYREX is a registered trademark of Corning, Inc.