Skip to Content
Merck
All Photos(1)

Key Documents

681075

Sigma-Aldrich

2-Chloro-6-methylpyridine-4-carbonyl chloride

97%

Synonym(s):

2-Chloro-6-methylisonicotinoyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5Cl2NO
CAS Number:
Molecular Weight:
190.03
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.5555

bp

80-85 °C/3-4 mmHg

density

1.361 g/mL at 25 °C

SMILES string

Cc1cc(cc(Cl)n1)C(Cl)=O

InChI

1S/C7H5Cl2NO/c1-4-2-5(7(9)11)3-6(8)10-4/h2-3H,1H3

InChI key

YHKLLTUNTUJQSB-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service