Skip to Content
Merck
All Photos(1)

Key Documents

197912

Sigma-Aldrich

2-Aminoimidazole sulfate

98%

Synonym(s):

2-Imidazoleamine sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5N3 · 0.5H2SO4
CAS Number:
Molecular Weight:
132.13
Beilstein:
4726750
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

270 °C (dec.) (lit.)

SMILES string

OS(O)(=O)=O.Nc1ncc[nH]1.Nc2ncc[nH]2

InChI

1S/2C3H5N3.H2O4S/c2*4-3-5-1-2-6-3;1-5(2,3)4/h2*1-2H,(H3,4,5,6);(H2,1,2,3,4)

InChI key

KUWRLKJYNASPQZ-UHFFFAOYSA-N

Application

2-Aminoimidazole sulfate was used in the synthesis of chlorohydrin.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A short synthesis of the marine bioactive metabolite (+/−) girolline.
Tetrahedron Letters, 39(44), 8085-8088 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service