Histology / Histopathology
Histology in Clinical Diagnostics
Histology is a field of biology and medicine dedicated to elucidating tissue structure, function, and disease states. The principle techniques of histology involve the use of various chemical stains to interrogate tissue samples ranging from single-celled organisms, plants, fungi, and animals, which are optimized for unique targets. Histopathology involves the clinical application of histological methods to examine diseased cells and tissues for diagnostic or prognostic analysis of various medical conditions like cancer and multi-organ diseases. It is also used to identify pathogens like bacteria, fungi, and parasites indicates the presence of some heavy metals and other toxins.
Featured Categories
We offer a complete selection of microscopy reagents, tools, and glassware for research and clinical lab analysis from Corning®, BRAND®, PELCO®, and other reliable specialty brands.
Milli-Q® systems offer innovative water purification technologies engineered to...
Despite the growing usage of antibody immunolabeling techniques in research and medicine today, clinical diagnostics remains largely centered on analysis based on well-established stains and dyes classified as routine, differentiating, and special. Routine stains like H&E are used as ubiquitous background stains. Differentiating stains include combinations of stains and counterstains to reveal more anatomical detail. Special stains are used in more detailed analyses, where closely related cell types need to be differentiated. Although some standard protocols are over 100 years old, histological staining is a vibrant and evolving field.
Staining in Histopathology
Numerous staining techniques were initially developed in an empirical way for analyzing tissue sections. The most specific staining mixtures were then developed for the staining and recognition of cell nuclei, cytoplasm, intra- and extra-cellular components.
Classical techniques, like H&E and Fuchsin staining, are adequate for most diagnoses (90-95%) however, some diagnosis require additional differential staining methods. These special methods (e.g., Wright-Giemsa, Schiff staining) enable the evaluation of additional morphological criteria and functional properties, ultimately making the final diagnosis more reliable. Additional techniques are now used to augment histopathologic analysis including immunohistochemical methods, DNA hybridization, fluorescence in situ hybridization, PCR, and flow cytometry.
Importance of Standardized Histological Staining
Diagnostic cytology, histology, and immunohistochemistry rely upon the dye, stain, or immune probe binding to the key cell and/or tissue chemical structures, that indicate the cellular architecture or pathological state. Thus, tissue structure preservation and permeability are crucial for consistent and reliable staining. Staining effectiveness can be influenced by tissue fixation type and time, tissue thickness, temperature, and target accessibility. Histopathology laboratories typically utilize standardized fixation and staining protocols with careful reagent management to minimize artifacts, high background, and false positives. Further, analysis of the tissue by subsequent staining, immunoprobing, or molecular techniques depends on the initial preservation and handling of the samples with established protocols.
Visit our document search for data sheets, certificates and technical documentation.
Related Articles
- Immunohistochemistry (IHC) represents an indispensable tool in cancer diagnostics.
- Fungal stains remain an important tool in the histology laboratory's diagnostic arsenal for identifying infectious microorganisms.
- Reliable histology staining requires consistent quality pure water. See examples of GMS and H&E stains performed with various water contaminants and comparing tap vs Type 2 water.
- Nitroblue Tetrazolium (NBT) is used with the alkaline phosphatase substrate 5-Bromo- 4-Chloro-3-Indolyl Phosphate (BCIP) in western blotting and immunohistological staining procedures. These substrate systems produce an insoluble NBT diformazan end product that is blue to purple in color and can be observed visually.
- CsCl gradient centrifugation separates RNA from DNA; differential and density gradient centrifugation techniques explained.
- See All (5)
Related Protocols
- Proper laboratory glassware cleaning techniques are crucial for high-quality results in experiments and research.
- Assay protocol for the colorimetric detection of Hydroxyproline in biological samples using the Hydroxypoline assay kit.
- Alcian Blue - H&E - Metanil Yellow Stain for Diagnosing Barrett's Esophagus. Barrett's esophagus is a condition that can lead to adenocarcinoma.
- This troubleshooting guide addresses the most common sources of error observed when using Histopaque but is not meant to be a comprehensive list.
- Detergent-free procedure is recommended for nuclear protein preparation to avoid interference with labeling efficiency of extracted proteins.
- See All (5)
Find More Articles and Protocols
How Can We Help
In case of any questions, please submit a customer support request
or talk to our customer service team:
Email [email protected]
or call +1 (800) 244-1173
Additional Support
- Chromatogram Search
Use the Chromatogram Search to identify unknown compounds in your sample.
- Calculators & Apps
Web Toolbox - science research tools and resources for analytical chemistry, life science, chemical synthesis and materials science.
- Customer Support Request
Customer support including help with orders, products, accounts, and website technical issues.
- FAQ