Merck
HomeMicrobiological AnalysisEN ISO 11133 Microbiological Culture Media
EN ISO 11133 Microbiological Culture Media
Riche E (PhD)1, Corpinot M (PhD)1, Plouhinec J2, Flint T1, Fischer S (PhD)3, Gerten B3, Hemy JC1, Airaud A1
1Lab Water Solutions, Merck, Guyancourt, France, 2Assystem Care, Courbevoie, France, 3LS-OII-Q Quality Specialties and BioMonitoring, {hcompany], Darmstadt, Germany
Centrally purified water vs. an autonomous water purification system
EN ISO 11133 and the quality control of microbiological culture media
Microbiology testing is critical to guarantee the safety and quality of food. Laboratories performing these tests are required to deliver accurate and reliable results and meet regulatory requirements while under time pressure. Many tests and procedures depend upon culture media being capable of providing consistent and reproducible results.
Learn about the ISO 11133:2014 standard and the impact of water contaminants on microbiology culture media in this hour long webinar, Guidance to ISO 11133 Compliance: Water Sourcing for Microbiology Culture Media. Or, read further in the Application Note: ISO 11133 and Water for Culture Media
The EN ISO 11133 standard establishes the context that ensures the quality of culture media and specifies the requirements for media preparation when used for the microbiological analysis of food, animal feed and water.1 In order to be compliant with the standard, each element of the process must meet specific requirements.
Water can potentially affect culture media performance since it is the largest component of microbiological media by volume. The purpose of the present study was to prepare culture media and to test their performance according to EN ISO 11133 using water from two alternative sources.
Centrally purified water vs. water from a Milli-Q® IX system and culture media performance
This study investigated whether a modern water purification system, the Milli-Q® IX pure water system, installed in a microbiology laboratory could advantageously replace centrally produced purified water obtained from a distribution loop when preparing and testing microbiological media. Solid and liquid culture media for important pathogens and hygiene indicators were included, such as Listeria, Salmonella, Escherichia coli serogroup O157 and coliforms, as well as culture media for yeast, molds and lactic acid bacteria.
Water sources used to prepare culture media
Each medium was prepared in duplicate by dissolving dehydrated medium in water from two different sources:
  • Purified water readily available in the QC laboratory: The purified water was centrally produced by a combination of reverse osmosis (RO), ion exchange and a bactericidal UV lamp, and delivered via a distribution loop. The water conductivity remained below 2 µS/cm throughout the study and the microbial level was < 102 cfu/mL, as required by the EN ISO 11133 standard (Table 1).
  • Water from a Milli-Q® IX 7003 water purification system: This system was installed in the lab and directly connected to tap water. It combined several water purification technologies to deliver pure water: RO, Elix® electrodeionization and bactericidal UVC LED lamps. Freshly purified water was collected from the system’s E-POD® dispenser, which was fitted with a Millipak® 0.22 µm filter to ensure low microbial levels. This system is validated to deliver purified water with a conductivity < 0.2 µS/cm, typically 0.1 µS/cm at 25 °C (equivalent to a resistivity > 5 MΩ.cm, typically 10-15 MΩ.cm), and microbial level < 102 cfu/mL, and fits the requirements of the ISO 11133 standard (Table 1).
 EN ISO 11133Centrally purified water in the QC laboratoryWater from Milli-Q® IX system fitted with a 0.22 µm filter
Water purityFor the preparation of culture media, use only purified water, i.e. distilled, demineralized, deionized or produced by reverse osmosis, or of equivalent quality free from substances likely to inhibit or influence the growth of the microorganisms under the test conditions e.g. traces of chlorine, traces of ammonia and traces of metal ions.Water is purified by a combination of reverse osmosis, ion-exchange and bactericidal UV lamp.The system contains a combination of purification technologies: reverse osmosis, Elix® electrodeionization, bactericidal UVC LED, 0.22 µm filtration. Data obtained during the system’s validation process:
· Free chlorine < 0.02 ppm
· Ammonia ≤ 0.00003%
· Heavy metals ≤ 0.00001%
StorageThe purified water shall be stored in tightly closed containers made from an inert material (neutral glass, polyethylene, etc.) which shall be free from all inhibitory substances. It is however recommended that the water is used as soon as produced. Not applicableWater is stored in a polyethylene tank, which is protected against airborne contaminants by a vent filter. An ergonomic E-POD® water dispenser provides easy delivery of pure water.
Microbial contaminationMicrobial contamination should not exceed 103 cfu/mL and preferably be below 102 cfu /mLMicrobial contamination ≤ 102 cfu/mLData obtained from an independent laboratory during the system’s validation process: microbial contamination ≤ 102 cfu/mL  
ConductivityThe conductivity shall be no more than 25 μS/cm (equivalent to resistivity ≥ 0.04 MΩ cm), preferably below 5 μS/cm (grade 3 water, ISO 3696) at 25 °C. The conductivity should be checked before use.Conductivity typically ≤ 2 µS/cmSystem validated to deliver water with conductivity < 0.2 μS/cm; typically 0.1 μS/cm. The system is designed to meet or exceed requirements as described by EN ISO 3696, Grade 2 water. Last produced water resistivity is displayed on the instrument.  
Table 1. Compliance of the two types of water tested with EN ISO 11133 requirements.1
Culture media preparation
Seven different dehydrated culture media were selected (Table 2) to obtain representative results. All media are compliant with the corresponding EN ISO/FDA Bacteriological Analytical Manual (BAM)/USDA-FSIS/APHA and other standards and methods were involved in performance testing. The standards specifying the media and the ones specifying their relative performance testing are reported in Table 3, as well as the control stains, and the WDCM and ATCC® (American Type Culture Collection) numbers.
The media were prepared following manufacturers’ instructions. Specifically, the required mass of the dehydrated media was added to 500 mL of centrally purified water or Milli-Q® IX water. All media were dissolved, heated and, if required, autoclaved as needed according to manufacturer’s instructions.
The study was performed by the Life Science QC laboratory for microbiological products at Merck, Darmstadt, Germany. The lab is accredited by the German accreditation authority DAkkS as registered test laboratory D-PL-15185-01-00 according to DIN EN ISO/IEC 170252 for the performance testing of media for microbiology according to DIN EN ISO 11133.
Culture mediumApplication of the culture mediumStandard specifying the medium
Listeria agar according to Ottaviani and AgostiFor the isolation and differentiation of Listeria monocytogenes and other Listeria spp. from food and animal feed, environmental samples in the area of food production and food handling, and other materials.ISO 11290, FDA-BAM, APHA
Violet red bile lactose (VRBL) agarFor the detection and colony counting of coliform bacteria from food and animal feed, water and other materials.ISO 4832, FDA-BAM, APHA
Malt extract agarFor the detection, isolation and enumeration of yeasts and molds in food products and other materials, and for their maintenance.-          
de Man, Rogosa and Sharpe (MRS) agarFor the isolation, enumeration and cultivation of Lactobacillus spp. and other mesophilic lactic acid bacteria from all types of materials.ISO 15214, APHA
Rappaport-Vassiliadis medium with soya (RVS) brothFor the selective enrichment of Salmonella spp. from food and animal feed, water and other materials.ISO 6579-1, ISO 19250
Brilliant green bile lactose (BRILA) brothFor the selective enrichment, enumeration and confirmation of Escherichia coli and other fecal coliform organisms from food and animal feed, water and other materials.ISO 4831, ISO 4832, FDA-BAM, APHA
Modified tryptone soya broth (mTSB) with novobiocin For the selective enrichment of Escherichia coli serogroup O157 in food and feed samples.ISO 16654
Table 2. Application of culture media used in this study and the standards specifying them.
Culture mediumStandard specifying the mediumStandard specifying the performance testingControl strains specified by the standardWDCM numbers specified by the standardWDCM numbers used in this study
Listeria agar according to Ottaviani and Agosti ISO 11290-1:2017
ISO 11290-2:2017
ISO 11290-1:2017
ISO 11290-2:2017
Listeria monocytogenes serovar 4b00021b00021
Listeria monocytogenes serovar 1/2a00109g00109
Escherichia coli0001200013d00012 00013
Enterococcus faecalis0000900087d00009 00087
Listeria innocua00017b00017
Violet red bile lactose (VRBL) agarISO 4832:2006EN ISO 11133:2014Escherichia coli0001200013g00012 00013
Enterococcus faecalis0000900087d00009 00087
Pseudomonas aeruginosa 00025b00025
Malt extract agar-----
de Man, Rogosa and Sharpe (MRS) agar ISO 15214:1998EN ISO 11133:2014Lactobacillus sakei 00015b00015
Lactococcus lactis00016b00016
Pediococcus pentosaceus00158g00158
Escherichia coli00012d 00013d00012 00013
Bacillus cereus00001g00001
Rappaport-Vassiliadis medium with soya (RVS) broth ISO 6579-1:2017ISO 6579-1:2017SalmonellaTyphimurium/ Enteritidis00030c,d 00031c,d00030 00031
Escherichia coli00012d 00013d00012 00013
Pseudomonas aeruginosa00025b00025
Enterococcus faecalis00009d 00087d00009 00087
Brilliant green bile lactose (BRILA) broth ISO 4831:2006
ISO 4832:2006
EN ISO 11133:2014 / Amd 2:2020Escherichia coli00012d 00013d 00090d 00179d00012 00013
Enterococcus faecalis00009d00087d 00176d00009 00087
Modified tryptone soya broth (mTSB) with novobiocin ISO 16654:2001----
Table 3. List of control strains for performance testing of culture media in the present study in accordance with the List of control strains for performance testing of culture media and reagents from published standards from ISO/TC 34/SC 9 & SC 5 (food microbiology)3 and ISO/TC 147/SC 4 (water microbiology)4.
bStrain to be used as minimum; cSome national restrictions and directions may require the use of a different serovar. Refer to national requirements relating to the choice of Salmonella serovars; dStrain free of choice; one of the positive strains has to be used as a minimum; gStrain optional. (Only the required footnotes are reported here).
Culture media evaluation: physical and chemical quality control
The seven culture media studied were prepared with water from the two alternative sources. Each prepared culture medium was assessed by visual inspection to ensure that it conformed to stated recommendations, e.g. appearance, color, homogeneity, gel consistency and pH.
Table 4 describes the physical parameters and the expected specifications, as well as the results obtained in both conditions. In each case, the media prepared with water from the Milli-Q® IX system matched the required specifications and gave similar outcomes as media prepared with the centrally produced purified water of the QC laboratory.
MediaPhysical parametersSpecificationsPurified water resultsMilli-Q® IX water results
Listeria agar according to Ottaviani and AgostiAppearance (clarity)slightly opalescent to opalescentopalescentopalescent
Appearance (color)yellowishyellowishyellowish
pH value (at 25 °C)7.0 – 7.47.17.1
Violet red bile lactose (VRBL) agar Appearance (clarity)clearclearclear
Appearance (color)redredred
pH value (at 25 °C)7.2 – 7.67.47.4
Solidification behavior (2 h at 45 °C)liquidliquidliquid
Malt extract agarAppearance (clarity)clear to slightly opalescentalmost clearalmost clear
Appearance (color)brownbrownbrown
pH value (at 25 °C)5.4 – 5.85.55.5
de Man, Rogosa and Sharpe (MRS) agar Appearance (clarity)clearclearclear
Appearance (color)brownbrownbrown
pH value (at 25 °C)5.6 – 5.85.65.6
Solidification behavior (2 h at 45 °C)liquidliquidliquid
Rappaport-Vassiliadis medium with soya (RVS) broth Appearance (clarity)clearclearclear
Appearance (color)dark bluedark bluedark blue
pH value (at 25 °C)5.0 – 5.45.45.4
Brilliant green bile lactose (BRILA) broth Appearance (clarity)clearclearclear
Appearance (color)greengreengreen
pH value (at 25 °C)7.0 – 7.47.27.2
Modified tryptone soya  broth (mTSB) with novobiocinAppearance (clarity)clearclearclear
Appearance (color)yellowish brownyellowish brownyellowish brown
pH value (at 25 °C)7.2 – 7.67.37.2
Table 4. Physical parameters and specifications of the seven culture media used in the study, as well as results obtained with either centrally purified water or water from a Milli-Q® IX system.
Performance testing of prepared microbiological culture media
Growth and inhibition were assessed by either quantitative or qualitative methods as described in the EN ISO 11133 standard.
  • Productivity is the level of recovery of a target microorganism from the culture medium under defined conditions.
  • Selectivity is the degree of inhibition of a non-target microorganism on or in a selective culture medium under defined conditions.
  • Specificity is the demonstration, under defined conditions, that non-target organisms, if a grow on the medium, do not show the same visual characteristics as target microorganisms.
1. Bacteria, yeasts and mold preparation and inoculation
Control strains were obtained from ATCC® and were selected according to the requirements of the EN ISO 11133 standard and/or other specific standards. They were diluted to the desired number of organisms for inoculation for testing on productivity, specificity and selectivity. Performance evaluation and interpretation of the results were obtained following the specifications given by EN ISO 11133.
2. Performance testing
  • Quantitative methods: Quantitative productivity for solid culture media was measured by quantitative methods as described in EN ISO 11133.
  • Qualitative methods: Productivity, specificity and selectivity were determined qualitatively using the methods as described by EN ISO 11133.
All the culture media prepared and tested met the required criteria for productivity, selectivity and specificity, when applicable. Quantitative productivity tests met the required criteria: productivity ratio (PR) ≥ 50% for selective media and PR ≥ 70% for non-selective media. Table 5 and Table 6 report the results obtained in the study. Qualitative and quantitative tests of the culture media led to results in accordance with the standard requirements with purified water used in the QC laboratory as well as with water delivered by the Milli-Q® IX system.
MediaFunction / Method of controlStrains testedSpecificationsPurified water resultsMilli-Q® IX water results
Listeria agar according to Ottaviani and AgostiProductivity / QuantitativeListeria monocytogenes ATCC® 13932™ [WDCM 00021]
Listeria monocytogenes ATCC® 35152™ [WDCM 00109]
Recovery rate ≥ 50%L. monocytogenes ATCC® 13932™: 117%, Blue green colonies with opaque halo
L. monocytogenes ATCC® 35152™: 74%, Blue green colonies with opaque halo
PASS
L. monocytogenes ATCC® 13932™: 124%, Blue green colonies with opaque halo
L. monocytogenes ATCC® 35152™: 83%, Blue green colonies with opaque halo
PASS
Selectivity / QualitativeEscherichia coli ATCC® 8739™ [WDCM 00012]
Escherichia coli ATCC® 25952™ [WDCM 00013]
Enterococcus faecalis ATCC® 29212™ [WDCM 00087]
Enterococcus faecalis ATCC® 19433™ [WDCM 00009]
Total inhibitionTotal inhibition
PASS
Total inhibition
PASS
Specificity / QualitativeListeria innocua ATCC® 33090™ [WDCM 00017]No limit
Growth, blue-green colonies without opaque halo
PASS
Growth, blue-green colonies without opaque halo
PASS
Violet red bile lactose (VRBL) agar
Productivity / QuantitativeEscherichia coli ATCC® 8739™ [WDCM 00012]
Escherichia coli ATCC® 25922™ [WDCM 00013]
Enterobacter cloacae ATCC® 13047™ [WDCM 00083]*
Recovery rate ≥ 50%E. coli ATCC® 8739™: 82%
E. coli ATCC® 25922™: 105%
E. cloacae: 85%
All: purplish-red colonies with or without precipitation halo
PASS
E. coli ATCC® 8739™: 103%
E. coli ATCC® 25922™: 102%
E. cloacae: 93%
All: purplish-red colonies with or without precipitation halo
 PASS
Selectivity / QualitativeEnterococcus faecalis ATCC® 29212™ [WDCM 00087]
Enterococcus faecalis ATCC® 19433™ [WDCM 00009]
Total inhibitionTotal inhibition
PASS
Total inhibition
PASS
Specificity / QualitativePseudomonas aeruginosa ATCC® 27853™ [WDCM 00025]Growth, no limit; colorless to beige coloniesGood growth, colorless to beige colonies
PASS
Good growth, colorless to beige colonies
PASS
Malt extract agarProductivity / QuantitativeCandida albicans ATCC® 10231™ [WDCM 00054]
Saccharomyces cerevisiae ATCC® 9763™ [WDCM 00058]
Saccharomyces cerevisiae ATCC® 9080™
Rhodotoula mucilaginosa DSM 70403™
Recovery rate ≥ 70%C. albicans: 82%
S. cerevisiae ATCC® 9763™: 107%
S. cerevisiae ATCC® 9080™: 87%
R. mucilaginosa: 113%
PASS
C. albicans: 82%
S. cerevisiae ATCC® 9763™: 100%
S. cerevisiae ATCC® 9080™: 102%
R. mucilaginosa: 116%
PASS
Specificity / QualitativeGeotrichum candidum DSM 1240™
Penicillium commune ATCC® 10428™
Aspergillus brasiliensis ATCC® 16404™ [WDCM 00053]
Good to very good growthVery good growth
PASS
Very good growth
PASS
Trichophyton ajelloi ATCC® 28454Moderate to good growthGood growth
PASS
Good growth
PASS
de Man, Rogosa and Sharpe (MRS) agar
Productivity / QuantitativeLactobacillus acidophilus ATCC® 4356™ [WDCM 00098]*
Lactobacillus sakei ATCC® 15521™ [WDCM 00015]
Lactococcus lactis ATCC® 19435™ [WDCM 00016]
Pediococcus pentosaceus ATCC® 33316™ [WDCM 00158]
Pediococcus damnosus ATCC® 29358™ [WDCM 00022]*
Recovery rate ≥ 70%L. acidophilus: 96%
L. sakei: 106%
L. lactis: 97%
P. pentosaceus: 91%
P. damnosus: 102%
PASS
L. acidophilus: 106%
L. sakei: 100%
L. lactis: 99%
P. pentosaceus: 88%
P. damnosus: 108%
PASS
Selectivity / QualitativeEscherichia coli ATCC® 25922™ [WDCM 00013]
Escherichia coli ATCC® 8739™ [WDCM 00012]
Bacillus cereus ATCC® 11778™ [WDCM 00001]
Total inhibitionTotal inhibition
PASS
Total inhibition
PASS
Bifidobacterium bifidum ATCC® 11863™*Good growthGood growth
PASS
Good growth
PASS
Table 5. Results of the performance tests of agar-based media prepared with either centrally purified water or water from a Milli-Q® IX system.
*Additional strains used in the experiments of the study. Strains tested for malt extract agar were chosen by the QC laboratory.
MediaFunction / Method of controlStrains testedSpecificationsPurified water resultsMilli-Q® IX water results
Rappaport-Vassiliadis medium with soya (RVS) brothProductivity / Qualitative Salmonella Typhimurium ATCC® 14028™ [WDCM 00031]+ Escherichia coli ATCC® 25922™ [WDCM 00013]+ Pseudomonas aeruginosa ATCC® 27853™ [WDCM 00025]> 10 colonies with black center on XLD agar> 10 colonies with black center on XLD agar
PASS 
> 10 colonies with black center on XLD agar
PASS
Salmonella Enteritidis ATCC® 13076™ [WDCM 00030]+ Escherichia coli ATCC® 8739™ [WDCM 00012]+ Pseudomonas aeruginosa ATCC® 27853™ [WDCM 00025]> 10 colonies with black center on XLD agar> 10 colonies with black center on XLD agar
PASS
> 10 colonies with black center on XLD agar
PASS
Selectivity / QualitativeEscherichia coli ATCC® 8739™ [WDCM 00012] Escherichia coli ATCC® 25922™ [WDCM 00013]Partial inhibition ≤ 100 colonies on Tryptic Soy Agar (TSA)For both, partial inhibition: ≤ 100 colonies on TSA;
PASS
For both, partial inhibition: ≤ 100 colonies on TSA
PASS
Enterococcus faecalis ATCC® 29212™ [WDCM 00087] Enterococcus faecalis ATCC® 19433™ [WDCM 00009]< 10 colonies on Tryptic Soy Agar (TSA)For both, < 10 colonies on TSA
PASS
For both, < 10 colonies on TSA
PASS
Brilliant green bile lactose (BRILA) brothProductivity / QualitativeEscherichia coli ATCC® 25922™ [WDCM 00013] Escherichia coli ATCC® 8739™ [WDCM 00012] Citrobacter freundii ATCC® 43864™ [WDCM 00006]*Turbidity and gasFor all 3: turbidity and gas
PASS
For all 3: turbidity and gas
PASS
Selectivity / QualitativeEnterococcus faecalis ATCC® 29212™ [WDCM 00087] Enterococcus faecalis ATCC® 19433™ [WDCM 00009]Partial inhibition, no gas productionFor both, partial inhibition, no gas production)
PASS
For both, partial inhibition, no gas production
PASS
Staphylococcus aureus ATCC® 6538™ [WDCM 00032]* Bacillus cereus ATCC® 11778™ [WDCM 00001]*Total inhibition, no gas productionFor both, total inhibition, no gas production
PASS
For both, total inhibition, no gas production
PASS
Modified tryptone soya broth (mTSB) with novobiocinProductivity / QualitativeEscherichia coli ATCC® 35150™ + Staphylococcus aureus ATCC® 25923™ [WDCM 00034] Escherichia coli ATCC® 700728™ [WDCM 00014] + Staphylococcus aureus ATCC® 25923™ [WDCM 00034]> 10 yellow-brown colonies on CT-SMAC AgarFor all, > 10 yellow-brown colonies on CT-SMAC Agar
PASS
For all, > 10 yellow-brown colonies on CT-SMAC Agar
PASS
Escherichia coli ATCC® 25922™ [WDCM 00013]+ Staphylococcus aureus ATCC® 25923™> 10 pink colonies on CT-SMAC agar> 10 pink colonies on CT-SMAC agar
PASS
> 10 pink colonies on CT-SMAC agar
PASS
Selectivity / QualitativeStaphylococcus aureus ATCC® 25923™ [WDCM 00034]Total inhibition on Tryptic Soy Agar (TSA)Total inhibition on TSA
PASS
Total inhibition on TSA
PASS
Table 6. Results of the performance tests of broth prepared with either centrally purified water or water from a Milli-Q® IX system.
*Additional strains used in the experiments of the study. Strains tested for mTSB were chosen by the QC laboratory.
Selecting a water solution for media preparation
When preparing culture media, it is important to select the correct water quality since water may contain impurities that can affect media productivity or cause abnormalities, such as incorrect pH, wrong color or precipitation, as indicated in Annex H of the EN ISO 11133 standard. The water quality requirements of the standard are shown in Table 1. An additional note in the standard indicates that water which has been passed through an ion exchange resin (demineralized or deionized water) can have a very high microorganism content. It is therefore advised not to use water from these sources without first checking microbial content. The standard warns that highly contaminated demineralized water, even if sterilized by filtration, can still contain substances that are inhibitory for the growth of certain microorganisms.
Suitability of the Milli-Q® IX water purification system to prepare and performance test microbiological culture media
This study demonstrates that culture media prepared with water from a Milli-Q® IX system has equivalent characteristics and performance as media prepared with the centrally purified water routinely used in the accredited QC lab. This indicates that water from this system can be used with confidence when preparing media according to the EN ISO 11133 standard. The Milli-Q® system can be used to prepare media manually or in combination with innovative automated instruments, such as the ReadyStream® system which simplifies the process by preparing and dispensing preheated culture media.
Water purified by the Milli-Q® IX water purification system, fitted with a 0.22 µm final filter, is validated to deliver water meeting or exceeding the water quality requirements of the EN ISO 11133 standard (Table 1). Other standards and methods, such as the American Public Health Association (APHA) Standard Methods,5 make specific recommendations regarding water quality and purification technologies, to which the Milli-Q® IX system abides by its design and performance. As recommended, it includes a combination of technologies to produce pure water, resistivity monitoring, as well as a storage tank equipped with UV irradiation. The Milli-Q® IX system delivers water with a conductivity below 0.2 µS/cm, microbial levels below 102 cfu/mL, and free from traces of contaminants likely to inhibit the growth of microorganisms, such as chlorine, ammonia and metals. Since the system delivers water that suits EN ISO 11133 water quality requirements, it will help to ensure a lab’s regulatory compliance. However, it is important to also ensure that this water is well suited for the preparation of microbiological culture media.
Design of the Milli-Q® IX system ensures constant quality pure water
Using water of consistent quality to prepare culture media is an important factor in ensuring accurate and reproducible test results. The Milli-Q® IX system is designed to produce constant quality pure water and to safeguard stored water purity via several key features:
  • Water purified by the system is stored in a high-quality polyethylene tank, which does not release extractables into water
  • The tank is protected against airborne contaminants by a vent filter.
  • Reliably low microbial levels are ensured by automatic recirculation of the pure water over a UVC LED bactericidal lamp, as well as the 0.22 µm screen filter placed at the system’s point-of-delivery.
  • The Elix® electrodeionization module included in the system delivers constant water quality, especially regarding conductivity6
The low level of extractables from the storage tank, the low microbial level in the water, as well as consistently low water conductivity are important water quality parameters highlighted in the EN ISO 11133 standard. Having a reliable and independent source of freshly purified water prevents any risk of unexpected changes in water quality or punctual unavailability of water from a central loop due to manufacturing constraints or maintenance.
Benefits of the Milli-Q® IX water purification system to prepare and performance test microbiological culture media
Using a Milli-Q® IX water purification system offers many benefits to labs performing microbiology testing.
  • According to the EN ISO 11133 standard, water conductivity (or its reciprocal, resistivity) should be checked before use. The Milli-Q® IX water system lets users accurately measure water resistivity on-line and displays it on the interface of the E-POD® dispenser, allowing a convenient monitoring solution. Checking water quality parameters is easy to perform via the intuitive touchscreen compatible with gloves.
  • The E-POD® dispenser allows time gain and precision as the flow rate can easily be adapted to the needs of the experiment, and the screen lets the user program a specific volume to be dispensed. Moreover, the dispenser brings flexibility as it can be used over the sink or rested on its arm to fill larger containers.
  • A foot pedal can be added to facilitate serial dispensing while freeing the user’s hands.
  • Regular system maintenance is easily done by any member of the lab, or can be delegated to a service engineer to ensure the system operates optimally.
  • The system stores all data related to water quality parameters, as well as information such as consumable changes and other service activities, thus providing data traceability and paperless data management. Easy-to-retrieve data can simplify the lab’s audit preparation. An online service is available to store all documentation and enable online service contract management.
  • For additional efficiency, the system can be used as a direct water source for the ReadyStream® system, to conveniently prepare large amounts (up to 100 L) of preheated media right where it is needed in only 15 minutes. The purified water delivered by the system can also be used for other tasks, such as glassware rinsing, and to feed autoclaves or glassware washers. This makes the Milli-Q® IX system a comprehensive solution for microbiology laboratories.
E-POD<sup>®</sup> dispenser of the Milli-Q<sup>®</sup> IX pure water system
The results of this study revealed that culture media prepared with water from the Milli-Q® IX water purification system performed similarly as the culture media prepared with the lab’s routinely used centrally purified water. This Milli-Q® water purification system enabled the lab to prepare media performing as required by the standard, while bringing additional benefits, such as ease of use, convenience, data traceability and autonomy. Combined, these features allow for efficient water system management, confidence in water purity and improved lab efficiency, which can keep scientists working at maximum productivity.
Related Products
Water Purification Solutions
Sorry, an unexpected error has occurred
Network error: Failed to fetch
Sorry, an unexpected error has occurred
Network error: Failed to fetch
References
1.
EN ISO International Standardization Organization. Microbiology of food, animal feed and water - Preparation, production, storage and performance testing of culture media + Amendment 1 + Amendment 2. EN ISO 11133:2014/Amd1:2018/ Amd2:2020.. [Internet]. Available from: https://www.iso.org/standard/70354
2.
ISO/IEC 17025:2017 - General requirements for the competence of testing and calibration laboratories. . [Internet]. Available from: http://santia.co.uk/wp-content/uploads/2017/10/P405-ISO-IEC-17025-2005-Standard.pdf
4.
ISO/TC 147 Water quality: Available from: https://www.iso.org/committee/52834.html
5.
2017. 9020 Quality assurance/Quality control; Standard Methods for the Examination of Water and Wastewater, 23rd Edition. [Internet]. Washington, D.C.: American Public Health Association. Available from: https://www.standardmethods.org/doi/10.2105/SMWW.2882.180
6.
Dimitrakopoulos T, Feuillas E, Darbouret D, Mabic S. Merck, Darmstadt, Germany. R&D Notebook 10.. Electrodeionization: technology and applications.