GC Analysis of 1-Aminoindan Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX™ G-DA
CONDITIONS
column
Astec CHIRALDEX G-DA, 30 m x 0.25 mm I.D., 0.12 μm (72033AST)
oven
150 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 24 psi
sample
peaks 1 & 2: 1-aminoindan enantiomers (N-TFA derivatives)
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC