Merck
  • Functional Langerinhigh-Expressing Langerhans-like Cells Can Arise from CD14highCD16- Human Blood Monocytes in Serum-Free Condition.
Functional Langerinhigh-Expressing Langerhans-like Cells Can Arise from CD14highCD16- Human Blood Monocytes in Serum-Free Condition.
Journal of immunology (Baltimore, Md. : 1950) (2016-03-27)
Gaëlle Picarda, Coraline Chéneau, Jean-Marc Humbert, Gaëlle Bériou, Paul Pilet, Jérôme Martin, Franck Duteille, Pierre Perrot, Frédérique Bellier-Waast, Michèle Heslan, Fabienne Haspot, Fabien Guillon, Regis Josien, Franck Albert Halary
ABSTRACT
Langerhans cells (LCs) are epithelial APCs that sense danger signals and in turn trigger specific immune responses. In steady-state, they participate in the maintenance of peripheral tolerance to self-antigens whereas under inflammation LCs efficiently trigger immune responses in secondary lymphoid organs. It has been demonstrated in mice that LC-deprived epithelia are rapidly replenished by short half-life langerin-expressing monocyte-derived LCs (MDLCs). These surrogate LCs are thought to be progressively replaced by langerin(high) LCs arising from self-renewing epithelial precursors of hematopoietic origin. How LCs arise from blood monocytes is not fully understood. Hence, we sought to characterize key factors that induce differentiation of langerin(high)-expressing monocyte-derived Langerhans-like cells. We identified GM-CSF and TGF-β1 as key cytokines to generate langerin(high)-expressing cells but only in serum-free conditions. These cells were shown to express the LC-specific TROP-2 and Axl surface markers and contained Birbeck granules. Surprisingly, E-cadherin was not spontaneously expressed by these cells but required a direct contact with keratinocytes to be stably induced. MDLCs induced stronger allogeneic T cell proliferations but released low amounts of inflammatory cytokines upon TLR stimulation compared with donor-paired monocyte-derived dendritic cells. Immature langerin(high) MDLCs were responsive to MIP-3β/CCL20 and CTAC/CCL27 chemokine stimulations. Finally, we demonstrated that those cells behaved as bona fide LCs when inserted in a three-dimensional rebuilt epithelium by becoming activated upon TLR or UV light stimulations. Collectively, these results prompt us to propose these langerin(high) MDLCs as a relevant model to address LC biology-related questions.
Sigma-Aldrich
Lipoprotein, low density from human plasma, lyophilized powder
Pricing and availability is not currently available.