Merck
Connexin-mimetic peptide Gap 27 decreases osteoclastic activity.
BMC musculoskeletal disorders (2001-12-19)
J Ilvesaro, P Tavi, J Tuukkanen
ABSTRACT
Bone remodelling is dependent on the balance between bone resorbing osteoclasts and bone forming osteoblasts. We have shown previously that osteoclasts contain gap-junctional protein connexin-43 and that a commonly used gap-junctional inhibitor, heptanol, can inhibit osteoclastic bone resorption. Since heptanol may also have some unspecific effect unrelated to gap-junctional inhibition we wanted to test the importance of gap-junctional communication to osteoclasts using a more specific inhibitor. A synthetic connexin-mimetic peptide, Gap 27, was used to evaluate the contribution of gap-junctional communication to osteoclastic bone resorption. We utilised the well-characterised pit-formation assay to study the effects of the specific gap-junctional inhibitor to the survival and activity of osteoclasts. Gap 27 caused a remarked decrease in the number of both TRAP-positive mononuclear and multinucleated rat osteoclasts cultured on bovine bone slices. The decrease in the cell survival seemed to be restricted to TRAP-positive cells, whereas the other cells of the culture model seemed unaffected. The activity of the remaining osteoclasts was found to be diminished by measuring the percentage of osteoclasts with actin rings of all TRAP-positive cells. In addition, the resorbed area in the treated cultures was greatly diminished. On the basis of these results we conclude that gap-junctional communication is essential for the action of bone resorbing osteoclasts and for proper remodelling for bone.
Sigma-Aldrich
Leukocyte Acid Phosphatase (TRAP) Kit, Select reagents packaged in gelatin capsules
Sigma-Aldrich
GAP-27, ≥97% (HPLC)