Merck
  • Knockdown of mutated H-Ras V12 expression induces chemosensitivity of hepatocellular carcinoma cells to cisplatin treatment in vitro and in nude mouse xenografts.
Knockdown of mutated H-Ras V12 expression induces chemosensitivity of hepatocellular carcinoma cells to cisplatin treatment in vitro and in nude mouse xenografts.
Oncology reports (2014-09-06)
Fanjie Meng, Bin Cao, Zengli Feng, Shunmao Ma, Haigang Wang, Yanshu Li, Hui Li
ABSTRACT
Ras mutations contribute to human cancer development. The present study assessed the Ras V12 mutation in hepatocellular carcinoma (HCC) cells and the role of its silencing in vitro and in nude mouse xenografts. HCC BEL7402 cells expressed mutations of V12 (Val/Gly) and wild-type H-Ras, whereas SMMC7721 cells only expressed wild-type H-Ras. The shRNA constructs specifically silenced mutated H-Ras expression in BEL7402 cells, but did not affect wild-type Ras expression in SMMC7721 cells. Silencing of mutated H-Ras expression reduced BEL7402 cell viability, induced apoptosis and arrested cells at the G0/G1 phase of the cell cycle. Expression of phospho-Akt was also significantly decreased, while several apoptotic proteins were also activated. Furthermore, sensitivity of stably H1-shRNA and H2-shRNA transfected BEL7402 cells to cisplatin treatment was increased by 7.19- and 5.39-fold, respectively, compared to that in the negative control cells, and apoptosis-related gene expression was also altered. Intraperitoneal administration of cisplatin led to a substantial reduction in HCC xenograft growth by 81 and 77% in the H1-shRNA and H2-shRNA transfected tumor xenografts, respectively, compared to a 37% reduction in mice bearing negative control tumor cells. These data demonstrate that knockdown of mutated H-Ras V12 expression induced chemosensitivity of HCC cells to cisplatin treatment in vitro and in vivo.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Hras1
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
USP
Transplatin, United States Pharmacopeia (USP) Reference Standard
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
MISSION® esiRNA, targeting human HRAS
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Cisplatin, European Pharmacopoeia (EP) Reference Standard