Merck
  • Sigma-1 receptor stimulation protects retinal ganglion cells from ischemia-like insult through the activation of extracellular-signal-regulated kinases 1/2.
Sigma-1 receptor stimulation protects retinal ganglion cells from ischemia-like insult through the activation of extracellular-signal-regulated kinases 1/2.
Experimental eye research (2014-10-12)
Brett H Mueller, Yong Park, Hai-Ying Ma, Adnan Dibas, Dorette Z Ellis, Abbot F Clark, Thomas Yorio
ABSTRACT
Sigma-1 receptor (σ-1) activation and mitogen-activated protein kinases (MAPKs) have been shown to protect retinal ganglion cells (RGCs) from cell death. The purpose of this study was to determine if σ-1 receptor stimulation with pentazocine could promote neuroprotection under conditions of an ischemia-like insult (oxygen glucose deprivation (OGD)) through the phosphorylation of extracellular signal regulated kinase (pERK)1/2. Primary RGCs were isolated from P3-P7 Sprague-Dawley rats and purified by sequential immunopanning using Thy1.1 antibodies. RGCs were cultured for 7 days before subjecting the cells to an OGD insult (0.5% oxygen in glucose-free medium) for 6 h. During the OGD, RGCs were treated with pentazocine (σ-1 receptor agonist) with or without BD 1047 (σ-1 receptor antagonist). In other experiments, primary RGCs were treated with pentazocine in the presence or absence of an MEK1/2 inhibitor, PD098059. Cell survival/death was assessed by staining with the calcein-AM/ethidium homodimer reagent. Levels of pERK1/2, total ERK1/2, and beta tubulin expression were determined by immunoblotting and immunofluorescence staining. RGCs subjected to OGD for 6 h induced 50% cell death in primary RGCs (p < 0.001) and inhibited pERK1/2 expression by 65% (p < 0.001). Cell death was attenuated when RGCs were treated with pentazocine under OGD (p < 0.001) and pERK1/2 expression was increased by 1.6 fold (p < 0.05) compared to OGD treated RGCs without pentazocine treatment. The co-treatment of PD098059 (MEK1/2 inhibitor) with pentazocine significantly abolished the protective effects of pentazocine on the RGCs during this OGD insult. Activation of the σ-1 receptor is a neuroprotective target that can protect RGCs from an ischemia-like insult. These results also established a direct relationship between σ-1 receptor stimulation and the neuroprotective effects of the ERK1/2 pathway in purified RGCs subjected to OGD. These findings suggest that activation of the σ-1 receptor may be a therapeutic target for neuroprotection particularly relevant to ocular neurodegenerative diseases that effect RGCs.
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
Forskolin, For use in molecular biology applications
Supelco
Forskolin, analytical standard
Sigma-Aldrich
(−)-Pentazocine, ≥98% (HPLC)
Sigma-Aldrich
(+)-Pentazocine, solid