Merck
  • Regulation of GLUT transporters by flavonoids in androgen-sensitive and -insensitive prostate cancer cells.
Regulation of GLUT transporters by flavonoids in androgen-sensitive and -insensitive prostate cancer cells.
Endocrinology (2014-06-17)
Pedro Gonzalez-Menendez, David Hevia, Aida Rodriguez-Garcia, Juan C Mayo, Rosa M Sainz
ABSTRACT
Cancer cells show different metabolic requirements from normal cells. In prostate cancer, particularly, glycolytic metabolism differs in androgen-responsive and nonresponsive cells. In addition, some natural compounds with antiproliferative activities are able to modify glucose entry into cells by either modulating glucose transporter (GLUT) expression or by altering glucose binding. The aim of this work was to study the regulation of some GLUTs (GLUT1 and GLUT4) in both androgen-sensitive (LNCaP) and -insensitive (PC-3) prostate cancer cells by 4 structurally different flavonoids (ie, genistein, phloretin, apigenin, and daidzein). Glucose uptake was measured using nonradiolabeled 2-deoxyglucose. The evaluation of protein levels as well as subcellular distribution of GLUT1/4 were analyzed by Western blot and immunocytochemistry, respectively. Androgen-insensitive LNCaP-R and androgen-sensitive PC-3-AR cells were used to study the effect of androgen signaling. Additionally, a docking simulation was employed to compare interactions between flavonoids and XylE, a bacterial homolog of GLUT1 to -4. Results show for the first time the presence of functionally relevant GLUT4 in prostate cancer cells. Furthermore, differences in GLUT1 and GLUT4 levels and glucose uptake were found, without differences on subcellular distribution, after incubation with flavonoids. Docking simulation showed that all compounds interact with the same location of transporters. More importantly, differences between androgen-sensitive and -insensitive prostate cancer cells were found in both GLUT protein levels and glucose uptake. Thus, phenotypic characteristics of prostate cancer cells are responsible for the different effects of these flavonoids in glucose uptake and in GLUT expression rather than their structural differences, with the most effective in reducing cell growth being the highest in modifying glucose uptake and GLUT levels.
Sigma-Aldrich
Resazurin sodium salt, certified by the Biological Stain Commission
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
Resazurin sodium salt, powder, BioReagent
Sigma-Aldrich
2-Deoxy-D-glucose, ≥99% (GC), crystalline
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), crystalline
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Trolamine, European Pharmacopoeia (EP) Reference Standard
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide phosphate sodium salt, pkg of 10 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide phosphate sodium salt, pkg of 5 mg (per vial)
Supelco
Daidzein, analytical standard
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Triethanolamine, puriss., meets analytical specification of NF, ≥99% (GC)
Sigma-Aldrich
Triethanolamine, ≥99.0% (GC)
Sigma-Aldrich
Triethanolamine, puriss. p.a., ≥99% (GC)
Sigma-Aldrich
Triethanolamine, reagent grade, 98%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide phosphate hydrate
Sigma-Aldrich
Daidzein, ≥98%, synthetic
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
Phloretin, ≥99%
Sigma-Aldrich
Triethanolamine, BioUltra, ≥99.5% (GC)