Merck
  • Discovery of L-755,507: a subnanomolar human beta 3 adrenergic receptor agonist.
Discovery of L-755,507: a subnanomolar human beta 3 adrenergic receptor agonist.
Bioorganic & medicinal chemistry letters (1999-01-01)
E R Parmee, H O Ok, M R Candelore, L Tota, L Deng, C D Strader, M J Wyvratt, M H Fisher, A E Weber
ABSTRACT
A study of 4-acylaminobenzenesulfonamides in a cloned human beta 3 adrenergic receptor assay resulted in the discovery of n-hexylurea, L-755,507 (22). This 0.43 nM beta 3 agonist, which is > 440-fold selective over both beta 1 and beta 2 binding, is among the most potent human beta 3 agonists reported to date.
Sigma-Aldrich
L755507, ≥98% (HPLC)