Merck
Review: ototoxic characteristics of platinum antitumor drugs.
Anatomical record (Hoboken, N.J. : 2007) (2012-10-10)
Dalian Ding, Brian L Allman, Richard Salvi
ABSTRACT
Cisplatin, carboplatin, nedaplatin, and oxaliplatin are widely used in contemporary oncology; however, their ototoxic and neurotoxic side effects are quite different as discussed in this review. Cisplatin is considered the most ototoxic, but despite its reputation, the magnitude of hair cell loss that occurs with a single, large drug bolus is limited and confined to the base of the cochlea. For all of these platinum compounds, a major factor limiting damage is drug uptake from stria vascularis into the cochlear fluids. Disrupting the blood-labyrinth barrier with diuretics or noise exposure enhances drug uptake and significantly increases the amount of damage. Combined treatment with ethacrynic acid (a loop diuretic) and cisplatin results in rapid apoptotic hair cell death characterized by upregulation of initiator caspase-8 and membrane death receptor, TRADD, followed by downstream executioners, caspase-3 and caspase-6. Unlike cisplatin, nedaplatin and oxaliplatin are highly neurotoxic when applied to cochlear cultures preferentially damaging auditory nerve fibers at low concentrations and hair cells at high concentrations. Carboplatin, considered far less ototoxic than cisplatin, is paradoxically highly toxic to chinchilla inner hair cells and type I spiral ganglion neurons; however, at high doses it also damages outer hair cells. Hair cell death from cisplatin and carboplatin is characterized in its early stages by upregulation of p53; blocking p53 expression with pifithrin-α prevents hair cell death. Major differences in the toxicity of these four platinum compounds may arise from several different metal transporters that selectively regulate the influx, efflux, and sequestration of these drugs.
Sigma-Aldrich
Platinum, foil, thickness 0.025 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum black, black, powder, ≤20 μm, ≥99.95% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.127 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum black, low bulk density, ≥99.9% trace metals basis
Sigma-Aldrich
Platinum black, ≥99.9% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 2.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.076 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Platinum, shot, ≤3 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Platinum, gauze, 100 mesh, 99.9% trace metals basis
Platinum, foil, not light tested, 50x50mm, thickness 0.004mm, as rolled, 99.95%
Platinum, foil, not light tested, 50x50mm, thickness 0.005mm, as rolled, 99.95%
Platinum, tube, 200mm, outside diameter 1.2mm, inside diameter 1.0mm, wall thickness 0.1mm, as drawn, 99.95%
Platinum, tube, 50mm, outside diameter 2.0mm, inside diameter 1.6mm, wall thickness 0.2mm, as drawn, 99.95%
Platinum, insulated wire, 2m, conductor diameter 0.025mm, insulation thickness 0.001mm, PTFE (polytetrafluoroethylene) insulation, 99.999%
Platinum, insulated wire, 0.5m, conductor diameter 0.05mm, insulation thickness 0.0075mm, polyester insulation, 99.99%
Platinum, insulated wire, 5m, conductor diameter 0.05mm, insulation thickness 0.009mm, PTFE (polytetrafluoroethylene) insulation, 99.99%
Platinum, tube, 200mm, outside diameter 0.95mm, inside diameter 0.60mm, wall thickness 0.175mm, as drawn, 99.95%
Platinum, insulated wire, 1m, conductor diameter 0.018mm, insulation thickness 0.002mm, polyester insulation, 99.99%
Platinum, insulated wire, 2m, conductor diameter 0.025mm, insulation thickness 0.005mm, polyester insulation, 99.99%
Platinum, rod, 50mm, diameter 1.25mm, 99.95%
Platinum, wire reel, 0.05m, diameter 0.25mm, as drawn, 99.998%
Platinum, tube, 500mm, outside diameter 1.2mm, inside diameter 1.0mm, wall thickness 0.1mm, as drawn, 99.95%
Platinum, microfoil, disks, 25mm, thinness 0.1μm, specific density 120μg/cm2, permanent mylar 3.5μm support, 99.99%
Platinum, tube, 100mm, outside diameter 1.0mm, inside diameter 0.8mm, wall thickness 0.1mm, as drawn, 99.95%
Platinum, rod, 10mm, diameter 4.0mm, 99.95%
Platinum, foil, 50x50mm, thickness 0.125mm, as rolled, 99.99+%