Merck
  • The effect of the irradiation wavelength on the processes sensitized by protoporphyrin IX dimethyl ester.
The effect of the irradiation wavelength on the processes sensitized by protoporphyrin IX dimethyl ester.
Radiation research (1997-10-27)
P Kubát, Z Zelinger, M Jirsa
ABSTRACT
The formation of triplet states of photosensitizers and singlet oxygen during reactions sensitized by protoporphyrin IX dimethyl ester (PPDME) and the products of its photo-oxidation in solution were studied by time-resolved spectroscopy. Irradiation with long-wavelength light (670 nm), which is absorbed by the products of photo-oxidation of PPDME, provides lower quantum yields of singlet oxygen than in sensitization with PPDME alone, which absorbs light with a wavelength of 630 nm. Spectroscopic measurements also confirmed the lower rate of sensitized photo-oxidation of bilirubin during irradiation with light with a wavelength of 670 nm.
Sigma-Aldrich
Protoporphyrin IX dimethyl ester, ~90% (HPLC)
Pricing and availability is not currently available.