Merck
Iodoquinol associated seizures and radiopacity.
Journal of toxicology. Clinical toxicology (1993-01-01)
A K Fisher, F G Walter, S Szabo
ABSTRACT
This case report describes seizures and encephalopathy in a nine year-old boy treated with iodoquinol for amebiasis. The toxicity of iodoquinol and other 8-hydroxyquinolines is discussed. The radiopacity of this medication, used worldwide for the treatment of diarrheal illnesses and dermatitis, may be diagnostically useful.
Sigma-Aldrich
5,7-Diiodo-8-hydroxyquinoline, 95%