Merck
  • Nucleoside influx and efflux in guinea-pig ventricular myocytes. Inhibition by analogues of lidoflazine.
Nucleoside influx and efflux in guinea-pig ventricular myocytes. Inhibition by analogues of lidoflazine.
Biochemical pharmacology (1994-08-30)
A R Conant, S M Jarvis
ABSTRACT
Adenosine influx and formycin B influx and efflux were characterized in guinea-pig ventricular myocytes at 22 degrees. Transport by both modes was saturable and inhibited by nitrobenzylthioinosine (NBMPR), indicating the presence of an equilibrative NBMPR-sensitive nucleoside transporter in the cardiomyocytes. The kinetic constants for influx and efflux of formycin B, a non-metabolized nucleoside, were similar, suggesting that the nucleoside transporter exhibits symmetrical kinetics (apparent Km 490 +/- 160 and 700 +/- 140 microM; Vmax 6.5 +/- 1.7 and 3.5 +/- 0.3 nmol/10(6) cells per min for influx and efflux, respectively). No evidence was found of either NBMPR-insensitive equilibrative nucleoside transport or sodium-dependent concentrative nucleoside transport. Inhibition of adenosine influx (apparent Km100 +/- 33 microM), by lidoflazine and the analogues mioflazine, soluflazine and R73-335, gave average Ki values of 730, 100, 64 and 2.9 nM, respectively. These compounds also inhibited formycin B efflux with a similar potency to that of adenosine influx. NBMPR-sensitive nucleoside transport was associated with high affinity binding of NBMPR (apparent Kd approximately 1 nM; 9.6 x 10(5) sites/cell). Specific binding of NBMPR was also inhibited by lidoflazine and its analogues. Mioflazine and soluflazine were 20-30-fold more potent at inhibiting NBMPR-sensitive nucleoside influx in guinea-pig erythrocytes than ventricular myocytes, indicating that the potency of some of the compounds studied is tissue dependent.
Sigma-Aldrich
Lidoflazine, ≥98% (HPLC), powder