Merck
  • Intermediary metabolism of pentachloronitrobenzene in the control and germ-free rat and rat with cannulated bile ducts.
Intermediary metabolism of pentachloronitrobenzene in the control and germ-free rat and rat with cannulated bile ducts.
Xenobiotica; the fate of foreign compounds in biological systems (1998-12-16)
G L Larsen, J K Huwe, J E Bakke
ABSTRACT
1. Nearly 70% of single oral doses of 14C-labelled pentachloronitrobenzene (PCNB) was excreted in bile within 24 h. 2. The characterized biliary metabolites of PCNB were either mercapturic acid pathway metabolites or catabolites thereof (thiols, methylthiols, S-glucuronides). 3. A major biliary metabolite was S-(aminotetrachlorophenyl)glutathione. 4. Conjugation with glutathione with subsequent catabolism to bis-methylthiotetrachlorobenzene was the major pathway in the control rat. 5. Germ-free experiments showed that only nitro- group displacement occurred, and no nitro- group reduction was detected.
Product Description
Supelco
Pentachloronitrobenzene solution, certified reference material, 5000 μg/mL in methanol
Supelco
Quintozene, PESTANAL®, analytical standard
Supelco
Pentachloroaniline, PESTANAL®, analytical standard
Sigma-Aldrich
Pentachloronitrobenzene, ≥94%