Merck
  • High resolution 11B NMR of magnesium diboride using cryogenic magic angle spinning.
High resolution 11B NMR of magnesium diboride using cryogenic magic angle spinning.
The Journal of chemical physics (2012-09-25)
Peter Beckett, Mark S Denning, Ivo Heinmaa, Mukesh C Dimri, Edward A Young, Raivo Stern, Marina Carravetta
ABSTRACT
Static and magic-angle spinning (11)B nuclear magnetic resonance (NMR) data at 4.7 T and 8.5 T have been obtained under cryogenic conditions on a diluted sample of magnesium diboride powder in the normal and superconducting state. The data provide accurate information on the magnetic shift and longitudinal relaxation time down to a temperature of 8 K, with a resolution improvement over the entire temperature range. The onset of superconductivity is unaffected by the sample rotation, as revealed by a steep variation of the magnetic shift just below the critical temperature.
Sigma-Aldrich
Magnesium boride, powder, −100 mesh, ≥99% trace metals basis