Merck
  • PPARγ regulates resistance vessel tone through a mechanism involving RGS5-mediated control of protein kinase C and BKCa channel activity.
PPARγ regulates resistance vessel tone through a mechanism involving RGS5-mediated control of protein kinase C and BKCa channel activity.
Circulation research (2012-09-11)
Pimonrat Ketsawatsomkron, Ramón A Lorca, Henry L Keen, Eric T Weatherford, Xuebo Liu, Christopher J Pelham, Justin L Grobe, Frank M Faraci, Sarah K England, Curt D Sigmund
ABSTRACT
Activation of peroxisome proliferator-activated receptor-γ (PPARγ) by thiazolidinediones lowers blood pressure, whereas PPARγ mutations cause hypertension. Previous studies suggest these effects may be mediated through the vasculature, but the underlying mechanisms remain unclear. To identify PPARγ mechanisms and transcriptional targets in vascular smooth muscle and their role in regulating resistance artery tone. We studied mesenteric artery (MA) from transgenic mice expressing dominant-negative (DN) mutant PPARγ driven by a smooth muscle cell-specific promoter. MA from transgenic mice exhibited a robust increase in myogenic tone. Patch clamp analysis revealed a reduced large conductance Ca(2+)-activated K(+) (BKCa) current in freshly dissociated smooth muscle cell from transgenic MA. Inhibition of protein kinase C corrected both enhanced myogenic constriction and impaired the large conductance Ca(2+)-activated K(+) channel function. Gene expression profiling revealed a marked loss of the regulator of G protein signaling 5 (RGS5) mRNA in transgenic MA, which was accompanied by a substantial increase in angiotensin II-induced constriction in MA. Small interfering RNA targeting RGS5 caused augmented myogenic tone in intact mesenteric arteries and increased activation of protein kinase C in smooth muscle cell cultures. PPARγ and PPARδ each bind to a PPAR response element close to the RGS5 promoter. RGS5 expression in nontransgenic MA was induced after activation of either PPARγ or PPARδ, an effect that was markedly blunted by DN PPARγ. We conclude that RGS5 in smooth muscle is a PPARγ and PPARδ target, which when activated blunts angiotensin II-mediated activation of protein kinase C, and preserves the large conductance Ca(2+)-activated K(+) channel activity, thus providing tight control of myogenic tone in the microcirculation.
Sigma-Aldrich
Tetraethylammonium chloride hydrate
Sigma-Aldrich
Tetraethylammonium iodide, 98%
Sigma-Aldrich
Calphostin C from Cladosporium cladosporioides, ≥90% (HPLC), powder
Sigma-Aldrich
Tetraethylammonium hydroxide solution, 20 wt. % in H2O
Sigma-Aldrich
Tetraethylammonium hydroxide solution, ~25% in methanol (~1.5 M)
Supelco
Tetraethylammonium hydroxide solution, ~1.0 M (CH3CH2)4NOH in H2O, electrochemical grade
Sigma-Aldrich
Tetraethylammonium hydroxide solution, 35 wt. % in H2O
Sigma-Aldrich
Tetraethylammonium chloride, ≥98% (titration)
Sigma-Aldrich
Tetraethylammonium bromide, ReagentPlus®, 99%
Sigma-Aldrich
Tetraethylammonium chloride, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
Tetraethylammonium bromide, reagent grade, 98%
Supelco
Tetraethylammonium chloride, for electrochemical analysis, ≥99.0%