Merck
  • The Pathological Effects of Melamine and Cyanuric Acid in the Diet of Walking Catfish (Clarius batrachus).
The Pathological Effects of Melamine and Cyanuric Acid in the Diet of Walking Catfish (Clarius batrachus).
Journal of comparative pathology (2012-02-10)
N Pirarat, T Katagiri, N Chansue, A Ponpornpisit, M Endo, M Maita
ABSTRACT
The toxicity of melamine and its analogue in man and animals has been reported widely. The aim of the present study was to examine the pathological effects of feeding melamine and cyanuric acid, separately or in combination, to walking catfish (Clarius batrachus). The catfish developed darkening of the skin as early as 3 days post feeding. Melamine-related crystals were distributed multifocally throughout the liver, kidney, heart, spleen and corpuscle of Stannius of fish fed melamine and cyanuric acid in combination. Oil red O staining and electron microscopy revealed that the melamine-related crystals had structure resembling that of plastic polymer crystals. Elevations in the serum concentrations of alanine transaminase, aspartate transaminase, creatinine and uric acid were related to the crystal-associated granulomatous inflammation in the liver and kidney of affected fish. None of the catfish died during the 2-week experiment. Melamine and cyanuric acid are therefore systemically toxic to fish in addition to causing renal crystal formation and renal damage as seen in man and animals. The finding of extrarenal crystals implies that the metabolism and biotransformation of these toxic compounds should be further investigated in aquatic animals.
Product Description
Supelco
Cyanuric acid, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Cyanuric acid, 98%
Pricing and availability is not currently available.