Merck
Engineering edgeless human skin with enhanced biomechanical properties.
Science advances (2023-01-28)
Alberto Pappalardo, David Alvarez Cespedes, Shuyang Fang, Abigail R Herschman, Eun Young Jeon, Kristin M Myers, Jeffrey W Kysar, Hasan Erbil Abaci
ABSTRACT
Despite the advancements in skin bioengineering, 3D skin constructs are still produced as flat tissues with open edges, disregarding the fully enclosed geometry of human skin. Therefore, they do not effectively cover anatomically complex body sites, e.g., hands. Here, we challenge the prevailing paradigm by engineering the skin as a fully enclosed 3D tissue that can be shaped after a body part and seamlessly transplanted as a biological clothing. Our wearable edgeless skin constructs (WESCs) show enhanced dermal extracellular matrix (ECM) deposition and mechanical properties compared to conventional constructs. WESCs display region-specific cell/ECM alignment, as well as physiologic anisotropic mechanical properties. WESCs replace the skin in full-thickness wounds of challenging body sites (e.g., mouse hindlimbs) with minimal suturing and shorter surgery time. This study provides a compelling technology that may substantially improve wound care and suggests that the recapitulation of the tissue macroanatomy can lead to enhanced biological function.
Sigma-Aldrich
Anti-Laminin alpha 5 Antibody, clone 4B12, clone 4B12, from mouse
Pricing and availability is not currently available.