Merck
  • Fast-to-slow shift of muscle fiber-type composition by dietary apple polyphenols in rats: Impact of the low-dose supplementation.
Fast-to-slow shift of muscle fiber-type composition by dietary apple polyphenols in rats: Impact of the low-dose supplementation.
Animal science journal = Nihon chikusan Gakkaiho (2016-07-16)
Wataru Mizunoya, Shinpei Okamoto, Hideo Miyahara, Mariko Akahoshi, Takahiro Suzuki, Mai-Khoi Q Do, Hideaki Ohtsubo, Yusuke Komiya, Mulan Qahar, Toshiaki Waga, Koichi Nakazato, Yoshihide Ikeuchi, Judy E Anderson, Ryuichi Tatsumi
ABSTRACT
Our previous studies demonstrated that an 8-week intake of 5% (w/w) apple polyphenol (APP) in the diet improves muscle endurance of young-adult rats. In order to identify a lower limit of the dietary contribution of APP to the effect, the experiments were designed for lower-dose supplementation (8-week feeding of 0.5% APP in AIN-93G diet) to 12-week-old male Sprague-Dawley rats. Results clearly showed that the 0.5% APP diet significantly up-regulates slower myosin-heavy-chain (MyHC) isoform ratios (IIx and IIa relative to total MyHC) and myoglobin expression in lower hind-limb muscles examined (P < 0.05). There was a trend to increased fatigue resistance detected from measurements of relative isometric plantar-flexion force torque generated by a stimulus train delivered to the tibial nerve (F(98, 1372) = 1.246, P = 0.0574). Importantly, there was no significant difference in the animal body-phenotypes or locomotor activity shown as total moving distance in light and dark periods. Therefore, the present study encourages the notion that even low APP-intake may increase the proportions of fatigue-resistant myofibers, and has promise as a strategy for modifying performance in human sports and improving function in age-related muscle atrophy.
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
Monoclonal Anti-Myoglobin antibody produced in mouse, clone MG-1, ascites fluid