Merck
  • Nuclear factor-kappaB regulates multiple steps of gliogenesis in the developing murine cerebral cortex.
Nuclear factor-kappaB regulates multiple steps of gliogenesis in the developing murine cerebral cortex.
Glia (2018-10-20)
Laurent Methot, Vincent Soubannier, Robert Hermann, Erin Campos, Sally Li, Stefano Stifani
ABSTRACT
Nuclear factor-kappaB (NF-κB) is activated in neural progenitor cells in the developing murine cerebral cortex during the neurogenic phase, when it acts to prevent premature neuronal differentiation. Here we show that NF-κB activation continues in mouse neocortical neural progenitor cells during the neurogenic-to-gliogenic switch. Blockade of endogenous NF-κB activity during neocortical gliogenesis leads to the formation of supernumerary committed gliogenic progenitors and premature glial cell differentiation. Conversely, forced NF-κB activation during the neocortical neurogenic-to-gliogenic transition causes delayed gliogenic commitment and decreased astroglial gene expression. NF-κB activation continues in neocortical gliogenic progenitors following commitment and is important to inhibit the differentiation of oligodendrocyte precursor cells and to maintain persistent expression of glial fibrillary acidic protein in maturing astrocytes. These results reveal a number of previously uncharacterized roles for NF-κB during different phases of neocortical gliogenesis and identify NF-κB as an inhibitor of early oligodendrocyte development in the cerebral cortex.
Sigma-Aldrich
Anti-NFIA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-S-100 (β-Subunit) antibody produced in mouse, clone SH-B1, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP) antibody produced in mouse, clone G-A-5, ascites fluid
Sigma-Aldrich
Anti-CNPase Antibody, clone 11-5B, clone 11-5B, Chemicon®, from mouse