Merck
  • Generation of hiPSC-derived low threshold mechanoreceptors containing axonal termini resembling bulbous sensory nerve endings and expressing Piezo1 and Piezo2.
Generation of hiPSC-derived low threshold mechanoreceptors containing axonal termini resembling bulbous sensory nerve endings and expressing Piezo1 and Piezo2.
Stem cell research (2021-10-05)
Shuyong Zhu, Nancy Stanslowsky, Jorge Fernández-Trillo, Tamrat M Mamo, Pengfei Yu, Norman Kalmbach, Birgit Ritter, Reto Eggenschwiler, Werner J D Ouwendijk, David Mzinza, Likai Tan, Andreas Leffler, Michael Spohn, Richard J P Brown, Kai A Kropp, Volkhard Kaever, Teng-Cheong Ha, Pratibha Narayanan, Adam Grundhoff, Reinhold Förster, Axel Schambach, Georges M G M Verjans, Manuela Schmidt, Andreas Kispert, Tobias Cantz, Ana Gomis, Florian Wegner, Abel Viejo-Borbolla
ABSTRACT
Somatosensory low threshold mechanoreceptors (LTMRs) sense innocuous mechanical forces, largely through specialized axon termini termed sensory nerve endings, where the mechanotransduction process initiates upon activation of mechanotransducers. In humans, a subset of sensory nerve endings is enlarged, forming bulb-like expansions, termed bulbous nerve endings. There is no in vitro human model to study these neuronal endings. Piezo2 is the main mechanotransducer found in LTMRs. Recent evidence shows that Piezo1, the other mechanotransducer considered absent in dorsal root ganglia (DRG), is expressed at low level in somatosensory neurons. We established a differentiation protocol to generate, from iPSC-derived neuronal precursor cells, human LTMR recapitulating bulbous sensory nerve endings and heterogeneous expression of Piezo1 and Piezo2. The derived neurons express LTMR-specific genes, convert mechanical stimuli into electrical signals and have specialized axon termini that morphologically resemble bulbous nerve endings. Piezo2 is concentrated within these enlarged axon termini. Some derived neurons express low level Piezo1, and a subset co-express both channels. Thus, we generated a unique, iPSCs-derived human model that can be used to investigate the physiology of bulbous sensory nerve endings, and the role of Piezo1 and 2 during mechanosensation.
Sigma-Aldrich
DAPT, ≥98% (HPLC), solid
Sigma-Aldrich
SU-5402, ≥98% (HPLC)
Sigma-Aldrich
N6,2′-O-Dibutyryladenosine 3′,5′-cyclic monophosphate sodium salt, ≥96% (HPLC), powder