Merck
  • DDR1 contributes to kidney inflammation and fibrosis by promoting the phosphorylation of BCR and STAT3.
DDR1 contributes to kidney inflammation and fibrosis by promoting the phosphorylation of BCR and STAT3.
JCI insight (2021-12-24)
Corina M Borza, Gema Bolas, Fabian Bock, Xiuqi Zhang, Favour C Akabogu, Ming-Zhi Zhang, Mark de Caestecker, Min Yang, Haichun Yang, Ethan Lee, Leslie Gewin, Agnes B Fogo, W Hayes McDonald, Roy Zent, Ambra Pozzi
ABSTRACT
Discoidin domain receptor 1 (DDR1), a receptor tyrosine kinase activated by collagen, contributes to chronic kidney disease. However, its role in acute kidney injury and subsequent development of kidney fibrosis is not clear. Thus, we performed a model of severe ischemia/reperfusion-induced acute kidney injury that progressed to kidney fibrosis in WT and Ddr1-null mice. We showed that Ddr1-null mice had reduced acute tubular injury, inflammation, and tubulointerstitial fibrosis with overall decreased renal monocyte chemoattractant protein (MCP-1) levels and STAT3 activation. We identified breakpoint cluster region (BCR) protein as a phosphorylated target of DDR1 that controls MCP-1 production in renal proximal tubule epithelial cells. DDR1-induced BCR phosphorylation or BCR downregulation increased MCP-1 secretion, suggesting that BCR negatively regulates the levels of MCP-1. Mechanistically, phosphorylation or downregulation of BCR increased β-catenin activity and in turn MCP-1 production. Finally, we showed that DDR1-mediated STAT3 activation was required to stimulate the secretion of TGF-β. Thus, DDR1 contributes to acute and chronic kidney injury by regulating BCR and STAT3 phosphorylation and in turn the production of MCP-1 and TGF-β. These findings identify DDR1 an attractive therapeutic target for ameliorating both proinflammatory and profibrotic signaling in kidney disease.
Sigma-Aldrich
Anti-CD167a/DDR1 Antibody, clone 5D5, clone 5D5, from mouse
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Roche
DNase I, from bovine pancreas
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
Interferon-γ human, IFN-gamma, recombinant, expressed in HEK 293 cells, suitable for cell culture, endotoxin tested