Merck
  • A mass spectrometry-based targeted assay for detection of SARS-CoV-2 antigen from clinical specimens.
A mass spectrometry-based targeted assay for detection of SARS-CoV-2 antigen from clinical specimens.
EBioMedicine (2021-07-07)
Santosh Renuse, Patrick M Vanderboom, Anthony D Maus, Jennifer V Kemp, Kari M Gurtner, Anil K Madugundu, Sandip Chavan, Jane A Peterson, Benjamin J Madden, Kiran K Mangalaparthi, Dong-Gi Mun, Smrita Singh, Benjamin R Kipp, Surendra Dasari, Ravinder J Singh, Stefan K Grebe, Akhilesh Pandey
ABSTRACT
The COVID-19 pandemic caused by severe acute respiratory syndrome-coronavirus 2 (SARS-CoV-2) has overwhelmed health systems worldwide and highlighted limitations of diagnostic testing. Several types of diagnostic tests including RT-PCR-based assays and antigen detection by lateral flow assays, each with their own strengths and weaknesses, have been developed and deployed in a short time. Here, we describe an immunoaffinity purification approach followed a by high resolution mass spectrometry-based targeted qualitative assay capable of detecting SARS-CoV-2 viral antigen from nasopharyngeal swab samples. Based on our discovery experiments using purified virus, recombinant viral protein and nasopharyngeal swab samples from COVID-19 positive patients, nucleocapsid protein was selected as a target antigen. We then developed an automated antibody capture-based workflow coupled to targeted high-field asymmetric waveform ion mobility spectrometry (FAIMS) - parallel reaction monitoring (PRM) assay on an Orbitrap Exploris 480 mass spectrometer. An ensemble machine learning-based model for determining COVID-19 positive samples was developed using fragment ion intensities from the PRM data. The optimized targeted assay, which was used to analyze 88 positive and 88 negative nasopharyngeal swab samples for validation, resulted in 98% (95% CI = 0.922-0.997) (86/88) sensitivity and 100% (95% CI = 0.958-1.000) (88/88) specificity using RT-PCR-based molecular testing as the reference method. Our results demonstrate that direct detection of infectious agents from clinical samples by tandem mass spectrometry-based assays have potential to be deployed as diagnostic assays in clinical laboratories, which has hitherto been limited to analysis of pure microbial cultures. This study was supported by DBT/Wellcome Trust India Alliance Margdarshi Fellowship grant IA/M/15/1/502023 awarded to AP and the generosity of Eric and Wendy Schmidt.
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, powder
Sigma-Aldrich
L-Alanine-1-13C, 99 atom % 13C