- An efficient and expeditious synthesis of di- and monosubstituted 2-aminoimidazoles.
An efficient and expeditious synthesis of di- and monosubstituted 2-aminoimidazoles.
Journal of combinatorial chemistry (2007-12-28)
Chai Hoon Soh, Wai Keung Chui, Yulin Lam
PMID18159933
ABSTRACT
A microwave-assisted protocol was developed for the construction of di- and monosubstituted 2-aminoimidazoles. The two-step reaction involves the synthesis of N-(1H-imidazol-2-yl)acetamides from readily available alpha-haloketones and N-acetylguanidine, followed by deacetylation. Significant rate enhancement was observed for both steps of the protocol, and the overall reaction time was shortened to 20 min compared to 48 h of the conventional procedures. A representative set of di- and monosubstituted 2-aminoimidazoles was prepared using commercially available parallel reactors.
MATERIALS