Merck
  • Extensive exploration of a novel rat model of Parkinson's disease using partial 6-hydroxydopamine lesion of dopaminergic neurons suggests new therapeutic approaches.
Extensive exploration of a novel rat model of Parkinson's disease using partial 6-hydroxydopamine lesion of dopaminergic neurons suggests new therapeutic approaches.
Synapse (New York, N.Y.) (2018-10-29)
Steven Vetel, Sophie Sérrière, Johnny Vercouillie, Jackie Vergote, Gabrielle Chicheri, Jean-Bernard Deloye, Frédéric Dollé, Sylvie Bodard, Claire Tronel, Lydie Nadal-Desbarats, Antoine Lefèvre, Patrick Emond, Sylvie Chalon
ABSTRACT
Parkinson's disease (PD) is characterized by the degeneration of dopaminergic (DA) neurons constituting the nigrostriatal pathway. Neuroinflammation, related to microglial activation, plays an important role in this process. Exploration of animal models of PD using neuroimaging modalities allows to better understand the pathophysiology of the disease. Here, we fully explored a moderate lesion model in the rat in which 6-hydroxydopamine was unilaterally delivered in three sites along the striatum. The degenerative process was assessed through in vivo Positron Emission Tomography (PET) imaging and in vitro autoradiographic quantitation of the striatal dopamine transporter (DAT) and immunostaining of tyrosine hydroxylase (TH). The microglial activation was studied through in vitro autoradiographic quantitation of the 18 kDa translocator protein (TSPO) in the striatum and CD11b staining in the SN. In addition, a targeted metabolomics exploration was performed in both these structures using mass spectrometry coupled to HPLC. Our results showed a reproducible decrease in the striatal DAT density associated with a reduction in the number of TH-positive cells in the SN and striatum, reflecting a robust moderate degeneration of nigrostriatal DA neurons. In addition, we observed strong microglia activation in both the striatum and SN ipsilateral to the lesion, highlighting that this moderate degeneration of DA neurons was associated with a marked neuroinflammation. Our metabolomics studies revealed alterations of specific metabolites and metabolic pathways such as carnitine, arginine/proline, and histidine metabolisms. These results bring new insights in the PD mechanism knowledge and new potential targets for future therapeutic strategies.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Anti-Integrin αM [CD11b] Antibody, clone OX-42, clone OX-42, Chemicon®, from mouse