Merck
  • Coupling of NMDA receptors and TRPM4 guides discovery of unconventional neuroprotectants.
Coupling of NMDA receptors and TRPM4 guides discovery of unconventional neuroprotectants.
Science (New York, N.Y.) (2020-10-10)
Jing Yan, C Peter Bengtson, Bettina Buchthal, Anna M Hagenston, Hilmar Bading
ABSTRACT
Excitotoxicity induced by NMDA receptors (NMDARs) is thought to be intimately linked to high intracellular calcium load. Unexpectedly, NMDAR-mediated toxicity can be eliminated without affecting NMDAR-induced calcium signals. Instead, excitotoxicity requires physical coupling of NMDARs to TRPM4. This interaction is mediated by intracellular domains located in the near-membrane portions of the receptors. Structure-based computational drug screening using the interaction interface of TRPM4 in complex with NMDARs identified small molecules that spare NMDAR-induced calcium signaling but disrupt the NMDAR/TRPM4 complex. These interaction interface inhibitors strongly reduce NMDA-triggered toxicity and mitochondrial dysfunction, abolish cyclic adenosine monophosphate-responsive element-binding protein (CREB) shutoff, boost gene induction, and reduce neuronal loss in mouse models of stroke and retinal degeneration. Recombinant or small-molecule NMDAR/TRPM4 interface inhibitors may mitigate currently untreatable human neurodegenerative diseases.
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
NMDAR/TRPM4 interface inhibitor C8 dihydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Diamide
Sigma-Aldrich
N-Methyl-D-aspartic acid, ≥98% (TLC), solid
Sigma-Aldrich
N6-Cyclopentyladenosine, solid
Sigma-Aldrich
Cyclopiazonic acid from Penicillium cyclopium, ≥98% (HPLC), powder
Sigma-Aldrich
NMDAR/TRPM4 interface inhibitor C19 dihydrochloride, ≥98% (HPLC)