Merck
  • Decrease of laminin-511 in the basement membrane due to photoaging reduces epidermal stem/progenitor cells.
Decrease of laminin-511 in the basement membrane due to photoaging reduces epidermal stem/progenitor cells.
Scientific reports (2020-07-30)
Shunsuke Iriyama, Masahito Yasuda, Saori Nishikawa, Eisuke Takai, Junichi Hosoi, Satoshi Amano
ABSTRACT
Daily sunlight exposure damages the epidermal basement membrane (BM) and disrupts epidermal homeostasis. Inter-follicular epidermal stem cells (IFE-SCs) regulate epidermal proliferation and differentiation, which supports epidermal homeostasis. Here, we examine how photoaging affects the function of IFE-SCs and we identify key components in their cellular environment (niche). We found that sun-exposed skin showed a decrease of MCSP-positive and β1-integrin-positive cells concomitantly with a decrease of laminin-511 at the dermal-epidermal junction (DEJ), as compared with sun-protected skin. Higher levels of laminin-511 were associated with not only increased efficiency of colony formation, but also higher expression levels of MCSP as well as other stem cell markers such as Lrig1, ITGB1, CD44, CD46, DLL1, and K15 in keratinocytes from skin of 12- to 62-year-old subjects. UVB exposure to cultured human skin impaired laminin-511 integrity at the dermal-epidermal junction and reduced MCSP-positive basal epidermal cells as well as K15-positive cells. Combined treatment with matrix metalloproteinase and heparanase inhibitors protected the integrity of laminin-511 and inhibited the reduction of MCSP-positive cells and K15-positive cells. These results suggest that photoaging may reduce the levels of MCSP-positive and K15-positive epidermal stem/progenitor cells in the epidermis via loss of laminin-511 at the dermal-epidermal junction.
Sigma-Aldrich
Anti-Laminin alpha 5 Antibody, clone 4B12, clone 4B12, from mouse
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Chondroitin Sulfate Proteoglycan Antibody, clone 9.2.27, clone 9.2.27, Chemicon®, from mouse