Merck
  • Cytocompatible poly(ethylene glycol)-co-polycarbonate hydrogels cross-linked by copper-free, strain-promoted click chemistry.
Cytocompatible poly(ethylene glycol)-co-polycarbonate hydrogels cross-linked by copper-free, strain-promoted click chemistry.
Chemistry, an Asian journal (2011-09-29)
Jianwen Xu, Tera M Filion, Fioleda Prifti, Jie Song
ABSTRACT
Strategies to encapsulate cells in cytocompatible three-dimensional hydrogels with tunable mechanical properties and degradability without harmful gelling conditions are highly desired for regenerative medicine applications. Here we reported a method for preparing poly(ethylene glycol)-co-polycarbonate hydrogels through copper-free, strain-promoted azide-alkyne cycloaddition (SPAAC) click chemistry. Hydrogels with varying mechanical properties were formed by "clicking" azido-functionalized poly(ethylene glycol)-co-polycarbonate macromers with dibenzocyclooctyne-functionalized poly(ethylene glycol) under physiological conditions within minutes. Bone marrow stromal cells encapsulated in these gels exhibited higher cellular viability than those encapsulated in photo-cross-linked poly(ethylene glycol) dimethacrylate. The precise control over the macromer compositions, cytocompatible SPAAC cross-linking, and the degradability of the polycarbonate segments make these hydrogels promising candidates for scaffold and stem cell assisted tissue repair and regeneration.
Sigma-Aldrich
Dibenzocyclooctyne-PEG4-acid, for Copper-free Click Chemistry
Pricing and availability is not currently available.
Sigma-Aldrich
Dibenzocyclooctyne-acid, 95%, storage temp.:-20°C
Pricing and availability is not currently available.