Merck
  • Heteropentameric cholera toxin B subunit chimeric molecules genetically fused to a vaccine antigen induce systemic and mucosal immune responses: a potential new strategy to target recombinant vaccine antigens to mucosal immune systems.
Heteropentameric cholera toxin B subunit chimeric molecules genetically fused to a vaccine antigen induce systemic and mucosal immune responses: a potential new strategy to target recombinant vaccine antigens to mucosal immune systems.
Infection and immunity (2005-08-23)
Tetsuya Harakuni, Hideki Sugawa, Ai Komesu, Masayuki Tadano, Takeshi Arakawa
ABSTRACT
Noninvasive mucosal vaccines are attractive alternatives to parenteral vaccines. Although the conjugation of vaccine antigens with the B subunit of cholera toxin (CTB) is one of the most promising strategies for vaccine delivery to mucosal immune systems, the molecule cannot tolerate large-protein fusion, as it severely impairs pentamerization and loses affinity for GM1-ganglioside. Here we report a new strategy, in which steric hindrance between CTB-antigen fusion subunits is significantly reduced through the integration of unfused CTB "molecular buffers" into the pentamer unit, making them more efficiently self-assemble into biologically active pentamers. In addition, the chimeric protein took a compact configuration, becoming small enough to be secreted, and one-step affinity-purified proteins, when administered through a mucosal route, induced specific immune responses in mice. Since our results are not dependent on the use of a particular expression system or vaccine antigen, this strategy could be broadly applicable to bacterial enterotoxin-based vaccine design.
Sigma-Aldrich
Phosphatase substrate, 40 mg tablets