Merck
  • Shielding Effect of Micelle for Highly Effective and Selective Monofluorination of Indoles in Water.
Shielding Effect of Micelle for Highly Effective and Selective Monofluorination of Indoles in Water.
ChemSusChem (2019-03-06)
Pranjal P Bora, Manisha Bihani, Scott Plummer, Fabrice Gallou, Sachin Handa
ABSTRACT
Highly selective direct monofluorination of indoles and arenes was developed through an approach that allows site-specific solubility of substrate and fluorine source in the micelle. This approach was highly selective for a broad range of substrates with excellent functional group tolerance. Differences in binding constant and solubility of indoles and arenes in the micelle allowed the fine-tuning of selectivity. Control experiments suggested a radical pathway and provided insight into the role of micelles of the environmentally benign amphiphile PS-750-M. Dynamic light scattering experiments strongly indicated the site-specific solubility of the substrate and fluorine source. The methodology was successfully adapted to gram scale, and the E-factor established from a recycle study indicated that the process is environmentally responsible and sustainable.
Sigma-Aldrich
PS-750-M
Pricing and availability is not currently available.
Sigma-Aldrich
PS-750-M, solution, 3 wt. % in H2O
Pricing and availability is not currently available.