Merck
Plasma and urinary oxalate and glycolate in healthy subjects.
Clinical chemistry (1993-01-01)
L Hagen, V R Walker, R A Sutton
ABSTRACT
High-performance ion chromatography (HPIC) has been widely used for oxalate analysis and, more recently, for glycolate analysis. We describe a procedure for sample preparation in which the plasma ultrafiltrate is acidified during harvesting with a cation-exchange resin, and the chloride is removed before the ion chromatography, which is performed with a newly developed AS10 column. The same ultrafiltrate sample is analyzed for glycolate. For plasma oxalate, the mean recovery of sample in eluted fractions was 95-96%, and intraassay CV was 6.2-8.1%. The reference interval (mean +/- 2 SD) for men was 0.8-3.2 mumol/L and for women, 1.0-2.6 mumol/L. For urinary oxalate, the reference interval for men was 175-560 mumol/day and for women, 107-432 mumol/day. For plasma glycolate, the mean analytical recovery was 96-98%, and the intra-assay CV was 2.4-6.2%. The reference interval for men was 1.9-7.5 mumol/L and for women, 1.4-7.4 mumol/L. For urinary glycolate, the reference interval for men was 0-1400 mumol/day and for women, 91-1001 mumol/day.
Sigma-Aldrich
100X GTPγS, 10mM